Assay Details
| Gene Symbol: LOC644145 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_003935.2 | 2 - 3 | 208 | 156 | ||
| GenBank mRNA | BC171876.1 | - | 2 - 3 | 256 | 156 | |
| DN831484.1 | - | 1 - 2 | 156 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | neuromedin U | NMU | -194.6kb | Anti-sense | Chr. 4 - 55595229 - 55636698 on NCBI Build GRCh38 | |
| Downstream | exocyst complex component 1 | EXOC1 |
BM-102, SEC3, SEC3L1, SEC3P |
16.4kb | Sense | Chr. 4 - 55853616 - 55905078 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04000323.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 122561 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 177534 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 8x60K | 49922 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 153537 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 760302 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 843051 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 699845 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 854729 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 176622 | AB085763.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 176622 | AK056934.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 176622 | BC013042.2 | genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 176622 | NM_145058.1 | refseq_mrna_human_NM |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 176622 | XM_001128421.1 | refseq_mrna_human_XM |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 176622 | XM_001134379.1 | refseq_mrna_human_XM |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 941104 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 136652 | AJ314646.2 | genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 136652 | AL359563.2 | genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 136652 | BC006240.2 | genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 136652 | BC047674.2 | genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 136652 | BC094803.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 136652 | NM_032932.3 | refseq_mrna_human_NM |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 11413 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 107449 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 281631 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 125244 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 96885 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3258349 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4286 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4285 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 43726 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P16688014 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P00977815 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P00977815 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3258349 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3258349 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23265744 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03111244 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23265777 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23265760 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03111245 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23265782 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23265763 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23265758 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23265726 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03111243 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03111257 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23265766 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03111223 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23265741 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23265753 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23265732 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23265731 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23265752 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23265754 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03111270 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03111260 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03111222 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23265747 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23265774 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03111249 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23265725 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23265779 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03111265 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23265734 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03111230 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23265781 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03111266 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23265728 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23265775 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23265771 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23265739 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03111250 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23265743 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23265767 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23265776 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23265778 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23265735 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23265779 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23265753 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03111230 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23265771 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03111265 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03111223 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23265778 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03111250 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23265735 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23265776 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23265739 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23265728 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23265726 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23265774 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23265758 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23265734 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23265754 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23265777 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23265760 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03111245 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23265732 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03111244 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23265781 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23265763 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23265752 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03111249 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23265741 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23265731 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23265725 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03111243 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03111266 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03111260 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23265743 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23265747 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23265775 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23265782 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03111270 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23265767 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03111222 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23265766 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23265744 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03111257 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03111266 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03111315 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03111278 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03111303 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03111284 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03111340 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03111332 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03111355 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03111373 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03111319 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03111239 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03111268 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03111229 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03111294 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03111270 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03111351 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03111272 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03111378 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15568686 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03111339 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03111343 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03111221 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03111260 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03111325 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03111255 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03111246 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03111219 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03111369 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03111317 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03111366 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03111243 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03111305 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03111365 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03111228 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03111235 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03111353 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03111251 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03111358 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03111371 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03111253 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03111321 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03111276 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03111297 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03111245 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03111376 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03111323 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03111362 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03111310 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03111241 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03111349 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03111224 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03111342 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03111262 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03111274 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03111348 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03111381 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03111217 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03111233 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03111335 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03111257 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03111345 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03111307 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03111291 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03111281 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03111327 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03111298 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03111223 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03111300 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03111288 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03111312 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03111248 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03111226 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03111264 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P16688014 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P16688069 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P00977803 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P00977815 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR04004545.hg.1 | TC04000323.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04004545.hg.1 | ENST00000504250.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04004545.hg.1 | NR_003935.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007570.hg.1 | ENST00000504250.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007570.hg.1 | ENST00000504250.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007570.hg.1 | ENST00000504250.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007570.hg.1 | ENST00000504250.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007570.hg.1 | ENST00000504250.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007570.hg.1 | NR_003935.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007570.hg.1 | NR_003935.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007570.hg.1 | NR_003935.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400151748.hg.1 | ENST00000504250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400151748.hg.1 | NR_003935.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400151748.hg.1 | ENST00000504250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400151748.hg.1 | NR_003935.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400151748.hg.1 | TC0400007570.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400007570.hg.1 | ENST00000504250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007570.hg.1 | ENST00000504250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007570.hg.1 | ENST00000504250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007570.hg.1 | ENST00000504250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007570.hg.1 | ENST00000504250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007570.hg.1 | NR_003935.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007570.hg.1 | NR_003935.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007570.hg.1 | NR_003935.2 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A007165 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A007165 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3237750 | NR_003935.2 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3237750 | DN831484.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3237750 | NR_003935.2 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3237750 | DN831484.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3237750 | NR_003935.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3237750 | DN831484.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3237750 | NR_003935.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3237750 | DN831484.1 | human_transcriptome |

LOC644145
Genomic Map