Assay Details
| Gene Symbol: IL17REL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001694.2 | NP_001001694.2 | 7 - 8 | 706 | 73 | |
| XM_011530689.2 | XP_011528991.1 | 7 - 8 | 1871 | 73 | ||
| XM_017028798.1 | XP_016884287.1 | 7 - 8 | 1871 | 73 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22000905.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR22015131.hg.1 | ENST00000389983.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015131.hg.1 | NM_001001694.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015131.hg.1 | TC22000905.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015127.hg.1 | ENST00000341280.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015127.hg.1 | ENST00000389983.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015127.hg.1 | NM_001001694.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015127.hg.1 | TC22000905.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009139.hg.1 | ENST00000341280.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009139.hg.1 | ENST00000341280.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009139.hg.1 | ENST00000341280.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009139.hg.1 | ENST00000341280.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009139.hg.1 | ENST00000341280.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009139.hg.1 | ENST00000341280.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009139.hg.1 | ENST00000341280.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009139.hg.1 | ENST00000341280.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009139.hg.1 | ENST00000341280.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009139.hg.1 | ENST00000341280.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009139.hg.1 | ENST00000341280.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009139.hg.1 | ENST00000341280.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009139.hg.1 | ENST00000341280.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009139.hg.1 | ENST00000341280.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009139.hg.1 | ENST00000341280.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009139.hg.1 | ENST00000341280.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009139.hg.1 | ENST00000389983.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009139.hg.1 | ENST00000389983.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009139.hg.1 | ENST00000389983.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009139.hg.1 | ENST00000389983.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009139.hg.1 | ENST00000389983.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009139.hg.1 | ENST00000389983.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009139.hg.1 | ENST00000389983.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009139.hg.1 | ENST00000389983.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009139.hg.1 | ENST00000389983.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009139.hg.1 | ENST00000389983.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009139.hg.1 | ENST00000389983.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009139.hg.1 | ENST00000389983.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009139.hg.1 | ENST00000389983.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009139.hg.1 | ENST00000389983.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009139.hg.1 | ENST00000389983.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009139.hg.1 | ENST00000389983.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009139.hg.1 | ENST00000389983.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009139.hg.1 | ENST00000389983.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009139.hg.1 | NM_001001694.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009139.hg.1 | NM_001001694.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009139.hg.1 | NM_001001694.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009139.hg.1 | NM_001001694.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009139.hg.1 | NM_001001694.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009139.hg.1 | NM_001001694.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009139.hg.1 | NM_001001694.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009139.hg.1 | NM_001001694.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009139.hg.1 | NM_001001694.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009139.hg.1 | NM_001001694.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009139.hg.1 | NM_001001694.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009139.hg.1 | NM_001001694.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009139.hg.1 | NM_001001694.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009139.hg.1 | NM_001001694.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009139.hg.1 | NM_001001694.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009139.hg.1 | NM_001001694.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009139.hg.1 | NM_001001694.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015128.hg.1 | ENST00000341280.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015128.hg.1 | ENST00000389983.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015128.hg.1 | NM_001001694.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015128.hg.1 | TC22000905.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015132.hg.1 | ENST00000389983.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015132.hg.1 | NM_001001694.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015132.hg.1 | TC22000905.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015125.hg.1 | ENST00000341280.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015125.hg.1 | ENST00000389983.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015125.hg.1 | NM_001001694.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015125.hg.1 | TC22000905.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015124.hg.1 | ENST00000341280.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015124.hg.1 | ENST00000389983.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015124.hg.1 | NM_001001694.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015124.hg.1 | TC22000905.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015126.hg.1 | ENST00000341280.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015126.hg.1 | ENST00000389983.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015126.hg.1 | NM_001001694.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015126.hg.1 | TC22000905.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015129.hg.1 | ENST00000341280.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015129.hg.1 | ENST00000389983.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015129.hg.1 | NM_001001694.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015129.hg.1 | TC22000905.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR2200166016.hg.1 | TC2200009139.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200166016.hg.1 | ENST00000341280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200166016.hg.1 | ENST00000389983.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200166016.hg.1 | NM_001001694.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200166016.hg.1 | ENST00000341280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200166016.hg.1 | ENST00000389983.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200166016.hg.1 | NM_001001694.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200166019.hg.1 | TC2200009139.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200166019.hg.1 | ENST00000341280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200166019.hg.1 | ENST00000389983.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200166019.hg.1 | NM_001001694.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200166019.hg.1 | ENST00000341280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200166019.hg.1 | ENST00000389983.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200166019.hg.1 | NM_001001694.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200166017.hg.1 | TC2200009139.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200166017.hg.1 | ENST00000341280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200166017.hg.1 | ENST00000389983.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200166017.hg.1 | NM_001001694.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200166017.hg.1 | ENST00000341280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200166017.hg.1 | ENST00000389983.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200166017.hg.1 | NM_001001694.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200166021.hg.1 | TC2200009139.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200166021.hg.1 | ENST00000341280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200166021.hg.1 | ENST00000389983.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200166021.hg.1 | NM_001001694.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200166021.hg.1 | ENST00000341280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200166021.hg.1 | ENST00000389983.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200166021.hg.1 | NM_001001694.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200166023.hg.1 | TC2200009139.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200166023.hg.1 | ENST00000389983.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200166023.hg.1 | NM_001001694.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200166023.hg.1 | ENST00000389983.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200166023.hg.1 | NM_001001694.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009139.hg.1 | ENST00000341280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009139.hg.1 | ENST00000341280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009139.hg.1 | ENST00000341280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009139.hg.1 | ENST00000341280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009139.hg.1 | ENST00000341280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009139.hg.1 | ENST00000341280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009139.hg.1 | ENST00000341280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009139.hg.1 | ENST00000341280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009139.hg.1 | ENST00000341280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009139.hg.1 | ENST00000341280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009139.hg.1 | ENST00000341280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009139.hg.1 | ENST00000341280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009139.hg.1 | ENST00000341280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009139.hg.1 | ENST00000341280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009139.hg.1 | ENST00000341280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009139.hg.1 | ENST00000341280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009139.hg.1 | ENST00000389983.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009139.hg.1 | ENST00000389983.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009139.hg.1 | ENST00000389983.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009139.hg.1 | ENST00000389983.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009139.hg.1 | ENST00000389983.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009139.hg.1 | ENST00000389983.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009139.hg.1 | ENST00000389983.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009139.hg.1 | ENST00000389983.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009139.hg.1 | ENST00000389983.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009139.hg.1 | ENST00000389983.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009139.hg.1 | ENST00000389983.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009139.hg.1 | ENST00000389983.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009139.hg.1 | ENST00000389983.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009139.hg.1 | ENST00000389983.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009139.hg.1 | ENST00000389983.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009139.hg.1 | ENST00000389983.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009139.hg.1 | ENST00000389983.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009139.hg.1 | ENST00000389983.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009139.hg.1 | NM_001001694.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009139.hg.1 | NM_001001694.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009139.hg.1 | NM_001001694.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009139.hg.1 | NM_001001694.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009139.hg.1 | NM_001001694.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009139.hg.1 | NM_001001694.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009139.hg.1 | NM_001001694.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009139.hg.1 | NM_001001694.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009139.hg.1 | NM_001001694.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009139.hg.1 | NM_001001694.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009139.hg.1 | NM_001001694.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009139.hg.1 | NM_001001694.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009139.hg.1 | NM_001001694.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009139.hg.1 | NM_001001694.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009139.hg.1 | NM_001001694.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009139.hg.1 | NM_001001694.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009139.hg.1 | NM_001001694.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200166018.hg.1 | TC2200009139.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200166018.hg.1 | ENST00000341280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200166018.hg.1 | ENST00000389983.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200166018.hg.1 | NM_001001694.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200166018.hg.1 | ENST00000341280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200166018.hg.1 | ENST00000389983.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200166018.hg.1 | NM_001001694.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200166024.hg.1 | TC2200009139.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200166024.hg.1 | ENST00000389983.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200166024.hg.1 | NM_001001694.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200166024.hg.1 | ENST00000389983.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200166024.hg.1 | NM_001001694.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200166020.hg.1 | TC2200009139.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200166020.hg.1 | ENST00000341280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200166020.hg.1 | ENST00000389983.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200166020.hg.1 | NM_001001694.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200166020.hg.1 | ENST00000341280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200166020.hg.1 | ENST00000389983.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200166020.hg.1 | NM_001001694.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200166015.hg.1 | TC2200009139.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200166015.hg.1 | ENST00000341280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200166015.hg.1 | ENST00000389983.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200166015.hg.1 | NM_001001694.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200166015.hg.1 | ENST00000341280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200166015.hg.1 | ENST00000389983.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200166015.hg.1 | NM_001001694.2 | Refseq |

IL17REL
Genomic Map