Assay Details
| Gene Symbol: GGH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003878.2 | NP_003869.1 | 6 - 7 | 888 | 164 | |
| GenBank mRNA | AK290450.1 | - | 6 - 7 | 632 | 164 | |
| BC025025.1 | - | 6 - 7 | 689 | 3924620 | 164 | |
| U55206.1 | - | 6 - 7 | 664 | 164 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08001269.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000518113.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000518113.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000518113.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000518113.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000518113.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000518113.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000518113.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000518466.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000518466.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000518466.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000518466.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000518466.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000518466.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000518966.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000518966.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000518966.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000518966.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000518966.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000518966.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000520609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000520609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000520609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000520609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000520609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000520609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000520609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000520609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000520609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000520609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000520609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000520609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000522852.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000522852.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000522852.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000523479.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000523479.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000523788.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000523788.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000523788.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000523788.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | ENST00000523788.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010630.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08016352.hg.1 | TC08001269.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08016352.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08016352.hg.1 | ENST00000518113.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08016352.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000518113.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000518113.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000518113.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000518113.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000518113.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000518113.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000518113.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000518466.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000518466.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000518466.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000518466.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000518466.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000518466.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000518966.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000518966.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000518966.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000518966.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000518966.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000518966.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000520609.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000520609.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000520609.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000520609.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000520609.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000520609.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000520609.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000520609.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000520609.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000520609.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000520609.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000520609.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000522852.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000522852.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000522852.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000523479.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000523479.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000523788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000523788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000523788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000523788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | ENST00000523788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010630.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800172171.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800172171.hg.1 | ENST00000518113.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800172171.hg.1 | ENST00000520609.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800172171.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800172171.hg.1 | ENST00000260118.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800172171.hg.1 | ENST00000518113.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800172171.hg.1 | ENST00000520609.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800172171.hg.1 | NM_003878.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800172171.hg.1 | TC0800010630.hg.1 | Clariom D TC |

GGH
Genomic Map