Assay Details
| Gene Symbol: CHCHD7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011669.2 | NP_001011669.1 | 2 - 3 | 328 | 133 | |
| NM_001317858.1 | NP_001304787.1 | 2 - 3 | 328 | 133 | ||
| NM_024300.4 | NP_077276.2 | 2 - 3 | 328 | 133 | ||
| GenBank mRNA | AK290088.1 | - | 2 - 3 | 133 | ||
| AY070434.1 | - | 2 - 3 | 176 | 133 | ||
| BC002546.2 | - | 2 - 3 | 167 | 3139026 | 133 | |
| BI911444.1 | - | 2 - 3 | 159 | 133 | ||
| HY130470.1 | - | 2 - 3 | 329 | 133 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08000387.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK290088.1,AY070434.1,BC002546.2,NM_001011669.2,NM_001317858.1,NM_024300.4 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR08005491.hg.1 | TC08000387.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08005491.hg.1 | ENST00000518801.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08005491.hg.1 | ENST00000519367.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08005491.hg.1 | ENST00000521524.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08005491.hg.1 | ENST00000521831.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08005491.hg.1 | NM_001011667.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08005491.hg.1 | NM_001011669.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08005491.hg.1 | NM_001011670.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08005492.hg.1 | TC08000387.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08005492.hg.1 | ENST00000303759.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08005492.hg.1 | ENST00000355315.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08005492.hg.1 | ENST00000396723.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08005492.hg.1 | ENST00000517933.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08005492.hg.1 | ENST00000518801.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08005492.hg.1 | ENST00000519367.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08005492.hg.1 | ENST00000521524.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08005492.hg.1 | ENST00000521831.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08005492.hg.1 | ENST00000523061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08005492.hg.1 | ENST00000523975.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08005492.hg.1 | NM_001011667.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08005492.hg.1 | NM_001011668.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08005492.hg.1 | NM_001011669.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08005492.hg.1 | NM_001011670.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08005492.hg.1 | NM_001011671.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08005492.hg.1 | NM_024300.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800152916.hg.1 | ENST00000518801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152916.hg.1 | ENST00000519367.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152916.hg.1 | ENST00000521524.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152916.hg.1 | ENST00000521831.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152916.hg.1 | ENST00000522166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152916.hg.1 | ENST00000523532.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152916.hg.1 | ENST00000523667.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152916.hg.1 | NM_001011667.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800152916.hg.1 | NM_001011669.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800152916.hg.1 | NM_001011670.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800152916.hg.1 | ENST00000518801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152916.hg.1 | ENST00000519367.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152916.hg.1 | ENST00000521524.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152916.hg.1 | ENST00000521831.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152916.hg.1 | ENST00000522166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152916.hg.1 | ENST00000523532.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152916.hg.1 | ENST00000523667.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152916.hg.1 | NM_001011667.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800152916.hg.1 | NM_001011669.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800152916.hg.1 | NM_001011670.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800152916.hg.1 | TC0800007697.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800152914.hg.1 | ENST00000303759.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152914.hg.1 | ENST00000355315.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152914.hg.1 | ENST00000396723.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152914.hg.1 | ENST00000517636.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152914.hg.1 | ENST00000517933.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152914.hg.1 | ENST00000518801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152914.hg.1 | ENST00000518944.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152914.hg.1 | ENST00000519367.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152914.hg.1 | ENST00000521524.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152914.hg.1 | ENST00000521831.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152914.hg.1 | ENST00000521982.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152914.hg.1 | ENST00000522166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152914.hg.1 | ENST00000522366.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152914.hg.1 | ENST00000523061.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152914.hg.1 | ENST00000523532.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152914.hg.1 | ENST00000523667.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152914.hg.1 | ENST00000523975.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152914.hg.1 | NM_001011667.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800152914.hg.1 | NM_001011668.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800152914.hg.1 | NM_001011669.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800152914.hg.1 | NM_001011670.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800152914.hg.1 | NM_001011671.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800152914.hg.1 | NM_024300.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800152914.hg.1 | ENST00000303759.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152914.hg.1 | ENST00000355315.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152914.hg.1 | ENST00000396723.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152914.hg.1 | ENST00000517636.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152914.hg.1 | ENST00000517933.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152914.hg.1 | ENST00000518801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152914.hg.1 | ENST00000518944.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152914.hg.1 | ENST00000519367.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152914.hg.1 | ENST00000521524.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152914.hg.1 | ENST00000521831.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152914.hg.1 | ENST00000521982.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152914.hg.1 | ENST00000522166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152914.hg.1 | ENST00000522366.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152914.hg.1 | ENST00000523061.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152914.hg.1 | ENST00000523532.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152914.hg.1 | ENST00000523667.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152914.hg.1 | ENST00000523975.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152914.hg.1 | NM_001011667.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800152914.hg.1 | NM_001011668.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800152914.hg.1 | NM_001011669.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800152914.hg.1 | NM_001011670.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800152914.hg.1 | NM_001011671.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800152914.hg.1 | NM_024300.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800152914.hg.1 | TC0800007697.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800152917.hg.1 | ENST00000303759.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152917.hg.1 | ENST00000355315.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152917.hg.1 | ENST00000396723.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152917.hg.1 | ENST00000517636.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152917.hg.1 | ENST00000517933.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152917.hg.1 | ENST00000518801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152917.hg.1 | ENST00000518944.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152917.hg.1 | ENST00000519367.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152917.hg.1 | ENST00000521524.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152917.hg.1 | ENST00000521831.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152917.hg.1 | ENST00000521982.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152917.hg.1 | ENST00000522166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152917.hg.1 | ENST00000523061.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152917.hg.1 | ENST00000523532.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152917.hg.1 | ENST00000523667.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152917.hg.1 | ENST00000523975.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152917.hg.1 | NM_001011667.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800152917.hg.1 | NM_001011668.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800152917.hg.1 | NM_001011669.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800152917.hg.1 | NM_001011670.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800152917.hg.1 | NM_001011671.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800152917.hg.1 | NM_024300.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800152917.hg.1 | ENST00000303759.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152917.hg.1 | ENST00000355315.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152917.hg.1 | ENST00000396723.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152917.hg.1 | ENST00000517636.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152917.hg.1 | ENST00000517933.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152917.hg.1 | ENST00000518801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152917.hg.1 | ENST00000518944.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152917.hg.1 | ENST00000519367.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152917.hg.1 | ENST00000521524.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152917.hg.1 | ENST00000521831.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152917.hg.1 | ENST00000521982.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152917.hg.1 | ENST00000522166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152917.hg.1 | ENST00000523061.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152917.hg.1 | ENST00000523532.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152917.hg.1 | ENST00000523667.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152917.hg.1 | ENST00000523975.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800152917.hg.1 | NM_001011667.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800152917.hg.1 | NM_001011668.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800152917.hg.1 | NM_001011669.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800152917.hg.1 | NM_001011670.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800152917.hg.1 | NM_001011671.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800152917.hg.1 | NM_024300.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800152917.hg.1 | TC0800007697.hg.1 | Clariom D TC |

CHCHD7
Genomic Map