| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10022262.hg.1 |
ENST00000355624.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10022262.hg.1 |
ENST00000369199.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10022262.hg.1 |
ENST00000483413.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10022262.hg.1 |
NM_014904.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10022262.hg.1 |
TC10001691.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011964.hg.1 |
ENST00000355624.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011964.hg.1 |
ENST00000355624.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011964.hg.1 |
ENST00000355624.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011964.hg.1 |
ENST00000355624.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011964.hg.1 |
ENST00000355624.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011964.hg.1 |
ENST00000355624.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011964.hg.1 |
ENST00000355624.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011964.hg.1 |
ENST00000355624.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011964.hg.1 |
ENST00000355624.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011964.hg.1 |
ENST00000355624.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011964.hg.1 |
ENST00000355624.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011964.hg.1 |
ENST00000355624.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011964.hg.1 |
ENST00000355624.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011964.hg.1 |
ENST00000369199.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011964.hg.1 |
ENST00000369199.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011964.hg.1 |
ENST00000369199.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011964.hg.1 |
ENST00000369199.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011964.hg.1 |
ENST00000369199.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011964.hg.1 |
ENST00000369199.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011964.hg.1 |
ENST00000369199.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011964.hg.1 |
ENST00000369199.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011964.hg.1 |
ENST00000369199.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011964.hg.1 |
ENST00000369199.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011964.hg.1 |
ENST00000369199.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011964.hg.1 |
ENST00000369199.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011964.hg.1 |
ENST00000369199.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011964.hg.1 |
ENST00000369199.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011964.hg.1 |
ENST00000476207.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011964.hg.1 |
ENST00000476207.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011964.hg.1 |
ENST00000476207.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011964.hg.1 |
ENST00000483413.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011964.hg.1 |
ENST00000483413.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011964.hg.1 |
ENST00000483413.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011964.hg.1 |
NM_014904.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011964.hg.1 |
NM_014904.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011964.hg.1 |
NM_014904.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011964.hg.1 |
NM_014904.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011964.hg.1 |
NM_014904.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011964.hg.1 |
NM_014904.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011964.hg.1 |
NM_014904.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011964.hg.1 |
NM_014904.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011964.hg.1 |
NM_014904.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011964.hg.1 |
NM_014904.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011964.hg.1 |
NM_014904.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011964.hg.1 |
NM_014904.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011964.hg.1 |
NM_014904.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10022261.hg.1 |
ENST00000355624.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10022261.hg.1 |
ENST00000369199.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10022261.hg.1 |
ENST00000483413.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10022261.hg.1 |
NM_014904.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10022261.hg.1 |
TC10001691.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1000181876.hg.1 |
ENST00000355624.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000181876.hg.1 |
ENST00000369199.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000181876.hg.1 |
ENST00000483413.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000181876.hg.1 |
NM_014904.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000181876.hg.1 |
ENST00000355624.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000181876.hg.1 |
ENST00000369199.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000181876.hg.1 |
ENST00000483413.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000181876.hg.1 |
NM_014904.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000181876.hg.1 |
TC1000011964.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1000181877.hg.1 |
ENST00000355624.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000181877.hg.1 |
ENST00000369199.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000181877.hg.1 |
ENST00000483413.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000181877.hg.1 |
NM_014904.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000181877.hg.1 |
ENST00000355624.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000181877.hg.1 |
ENST00000369199.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000181877.hg.1 |
ENST00000483413.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000181877.hg.1 |
NM_014904.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000181877.hg.1 |
TC1000011964.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1000011964.hg.1 |
ENST00000355624.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011964.hg.1 |
ENST00000355624.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011964.hg.1 |
ENST00000355624.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011964.hg.1 |
ENST00000355624.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011964.hg.1 |
ENST00000355624.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011964.hg.1 |
ENST00000355624.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011964.hg.1 |
ENST00000355624.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011964.hg.1 |
ENST00000355624.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011964.hg.1 |
ENST00000355624.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011964.hg.1 |
ENST00000355624.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011964.hg.1 |
ENST00000355624.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011964.hg.1 |
ENST00000355624.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011964.hg.1 |
ENST00000355624.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011964.hg.1 |
ENST00000369199.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011964.hg.1 |
ENST00000369199.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011964.hg.1 |
ENST00000369199.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011964.hg.1 |
ENST00000369199.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011964.hg.1 |
ENST00000369199.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011964.hg.1 |
ENST00000369199.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011964.hg.1 |
ENST00000369199.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011964.hg.1 |
ENST00000369199.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011964.hg.1 |
ENST00000369199.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011964.hg.1 |
ENST00000369199.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011964.hg.1 |
ENST00000369199.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011964.hg.1 |
ENST00000369199.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011964.hg.1 |
ENST00000369199.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011964.hg.1 |
ENST00000369199.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011964.hg.1 |
ENST00000476207.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011964.hg.1 |
ENST00000476207.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011964.hg.1 |
ENST00000476207.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011964.hg.1 |
ENST00000483413.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011964.hg.1 |
ENST00000483413.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011964.hg.1 |
ENST00000483413.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011964.hg.1 |
NM_014904.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011964.hg.1 |
NM_014904.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011964.hg.1 |
NM_014904.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011964.hg.1 |
NM_014904.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011964.hg.1 |
NM_014904.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011964.hg.1 |
NM_014904.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011964.hg.1 |
NM_014904.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011964.hg.1 |
NM_014904.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011964.hg.1 |
NM_014904.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011964.hg.1 |
NM_014904.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011964.hg.1 |
NM_014904.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011964.hg.1 |
NM_014904.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011964.hg.1 |
NM_014904.2 |
Refseq |