Assay Details
| Gene Symbol: NUDT14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318380.1 | NP_001305309.1 | 1 - 2 | 200 | 139 | |
| NM_177533.4 | NP_803877.2 | 1 - 2 | 200 | 139 | ||
| GenBank mRNA | AK128196.1 | - | 1 - 2 | 189 | 139 | |
| AK310449.1 | - | 1 - 2 | 177 | 139 | ||
| BC041584.1 | - | 1 - 2 | 162 | 4810207 | 139 | |
| DA249638.1 | - | 1 - 2 | 120 | 139 | ||
| DA409636.1 | - | 1 - 2 | 183 | 140 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14002334.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK128196.1,BC041584.1,NM_001318380.1,NM_177533.4 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR14017558.hg.1 | TC14002334.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR14017558.hg.1 | ENST00000392568.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14017558.hg.1 | ENST00000535832.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14017558.hg.1 | NM_177533.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14017552.hg.1 | TC14002334.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR14017552.hg.1 | ENST00000392568.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14017552.hg.1 | ENST00000535832.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14017552.hg.1 | NM_177533.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14017550.hg.1 | TC14002334.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR14017550.hg.1 | ENST00000392568.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14017550.hg.1 | ENST00000535832.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14017550.hg.1 | NM_177533.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14017551.hg.1 | TC14002334.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR14017551.hg.1 | ENST00000392568.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14017551.hg.1 | ENST00000535832.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14017551.hg.1 | NM_177533.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000339418.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000339418.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000339418.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000339418.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000339418.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000339418.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000339418.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000339418.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000392568.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000392568.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000392568.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000392568.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000392568.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000392568.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000392568.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000392568.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000392568.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000392568.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000546553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000546553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000546553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000546553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000546553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000546553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000546553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000546553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000550002.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000550002.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000550002.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000550002.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000550002.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000550912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000550912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000550912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000551024.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000551024.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000551024.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000551024.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000551024.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000551024.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000551024.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000551024.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000551024.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000551024.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | NM_177533.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | NM_177533.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | NM_177533.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | NM_177533.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | NM_177533.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | NM_177533.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | NM_177533.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | NM_177533.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | NM_177533.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | NM_177533.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400183537.hg.1 | ENST00000339418.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183537.hg.1 | ENST00000392568.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183537.hg.1 | ENST00000546553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183537.hg.1 | ENST00000551024.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183537.hg.1 | NM_177533.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400183537.hg.1 | ENST00000339418.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183537.hg.1 | ENST00000392568.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183537.hg.1 | ENST00000546553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183537.hg.1 | ENST00000551024.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183537.hg.1 | NM_177533.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400183537.hg.1 | TC1400010797.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000339418.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000339418.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000339418.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000339418.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000339418.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000339418.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000339418.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000339418.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000392568.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000392568.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000392568.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000392568.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000392568.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000392568.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000392568.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000392568.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000392568.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000392568.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000546553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000546553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000546553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000546553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000546553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000546553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000546553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000546553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000550002.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000550002.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000550002.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000550002.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000550002.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000550912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000550912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000550912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000551024.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000551024.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000551024.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000551024.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000551024.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000551024.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000551024.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000551024.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000551024.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000551024.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | NM_177533.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | NM_177533.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | NM_177533.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | NM_177533.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | NM_177533.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | NM_177533.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | NM_177533.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | NM_177533.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | NM_177533.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | NM_177533.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400183534.hg.1 | ENST00000339418.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183534.hg.1 | ENST00000392568.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183534.hg.1 | ENST00000546553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183534.hg.1 | ENST00000550002.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183534.hg.1 | ENST00000551024.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183534.hg.1 | NM_177533.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400183534.hg.1 | ENST00000339418.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183534.hg.1 | ENST00000392568.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183534.hg.1 | ENST00000546553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183534.hg.1 | ENST00000550002.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183534.hg.1 | ENST00000551024.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183534.hg.1 | NM_177533.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400183534.hg.1 | TC1400010797.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400183536.hg.1 | ENST00000339418.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183536.hg.1 | ENST00000392568.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183536.hg.1 | ENST00000546553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183536.hg.1 | ENST00000551024.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183536.hg.1 | NM_177533.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400183536.hg.1 | ENST00000339418.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183536.hg.1 | ENST00000392568.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183536.hg.1 | ENST00000546553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183536.hg.1 | ENST00000551024.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183536.hg.1 | NM_177533.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400183536.hg.1 | TC1400010797.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400183538.hg.1 | ENST00000392568.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183538.hg.1 | ENST00000546553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183538.hg.1 | ENST00000551024.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183538.hg.1 | NM_177533.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400183538.hg.1 | ENST00000392568.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183538.hg.1 | ENST00000546553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183538.hg.1 | ENST00000551024.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183538.hg.1 | NM_177533.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400183538.hg.1 | TC1400010797.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 104588 | - | - |

NUDT14
Genomic Map