Assay Details
| Gene Symbol: ANXA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320880.1 | NP_001307809.1 | 4 - 5 | 451 | 118 | |
| NM_004034.3 | NP_004025.1 | 5 - 6 | 506 | 118 | ||
| XM_017016162.1 | XP_016871651.1 | 5 - 6 | 542 | 118 | ||
| GenBank mRNA | AK295319.1 | - | 4 - 5 | 317 | 118 | |
| AK312447.1 | - | 5 - 6 | 479 | 118 | ||
| DA131184.1 | - | 4 - 5 | 424 | 118 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10001408.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
cellular calcium ion homeostasis
autophagy
hemostasis
cell proliferation
regulation of cell shape
response to salt stress
cellular water homeostasis
negative regulation of gene expression
response to organic cyclic compound
epithelial cell differentiation
social behavior
response to calcium ion
membrane fusion
integrin binding
calcium ion binding
protein binding
calcium-dependent phospholipid binding
poly(A) RNA binding
calcium-dependent protein binding
cellular calcium ion homeostasis
autophagy
hemostasis
cell proliferation
regulation of cell shape
response to salt stress
cellular water homeostasis
negative regulation of gene expression
response to organic cyclic compound
epithelial cell differentiation
social behavior
response to calcium ion
membrane fusion
integrin binding
calcium ion binding
protein binding
calcium-dependent phospholipid binding
poly(A) RNA binding
calcium-dependent protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR10017756.hg.1 | ENST00000372919.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10017756.hg.1 | NM_004034.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10017756.hg.1 | TC10001408.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | ENST00000372919.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | ENST00000372919.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | ENST00000372919.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | ENST00000372919.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | ENST00000372919.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | ENST00000372919.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | ENST00000372919.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | ENST00000372919.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | ENST00000372919.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | ENST00000372919.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | ENST00000372919.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | ENST00000372919.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | ENST00000372919.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | ENST00000372919.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | ENST00000372919.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | ENST00000372919.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | ENST00000372919.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | ENST00000372919.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | ENST00000372919.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | ENST00000372919.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | ENST00000372921.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | ENST00000372921.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | ENST00000372921.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | ENST00000372921.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | ENST00000372921.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | ENST00000372921.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | ENST00000372921.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | ENST00000372921.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | ENST00000372921.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | ENST00000372921.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | ENST00000372921.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | ENST00000372921.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | ENST00000372921.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | ENST00000372921.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | ENST00000372921.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | ENST00000372921.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | ENST00000372921.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | ENST00000372921.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | ENST00000372921.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | ENST00000372921.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | ENST00000394847.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | ENST00000394847.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | ENST00000394847.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | ENST00000394847.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | ENST00000394847.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | ENST00000394847.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | ENST00000463788.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | ENST00000463788.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | ENST00000463788.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | ENST00000463788.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | ENST00000463788.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | ENST00000492380.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | ENST00000492380.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | ENST00000492380.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | ENST00000492380.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | ENST00000492380.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | ENST00000492380.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | ENST00000492380.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | ENST00000492380.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | ENST00000492380.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | NM_001156.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | NM_001156.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | NM_001156.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | NM_001156.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | NM_001156.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | NM_001156.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | NM_001156.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | NM_001156.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | NM_001156.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | NM_001156.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | NM_001156.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | NM_001156.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | NM_001156.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | NM_001156.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | NM_001156.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | NM_001156.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | NM_001156.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | NM_001156.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | NM_001156.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | NM_004034.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | NM_004034.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | NM_004034.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | NM_004034.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | NM_004034.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | NM_004034.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | NM_004034.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | NM_004034.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | NM_004034.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | NM_004034.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | NM_004034.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | NM_004034.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | NM_004034.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | NM_004034.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | NM_004034.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | NM_004034.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | NM_004034.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | NM_004034.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | NM_004034.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011020.hg.1 | NM_004034.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | ENST00000372919.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | ENST00000372919.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | ENST00000372919.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | ENST00000372919.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | ENST00000372919.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | ENST00000372919.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | ENST00000372919.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | ENST00000372919.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | ENST00000372919.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | ENST00000372919.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | ENST00000372919.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | ENST00000372919.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | ENST00000372919.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | ENST00000372919.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | ENST00000372919.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | ENST00000372919.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | ENST00000372919.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | ENST00000372919.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | ENST00000372919.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | ENST00000372919.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | ENST00000372921.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | ENST00000372921.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | ENST00000372921.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | ENST00000372921.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | ENST00000372921.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | ENST00000372921.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | ENST00000372921.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | ENST00000372921.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | ENST00000372921.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | ENST00000372921.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | ENST00000372921.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | ENST00000372921.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | ENST00000372921.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | ENST00000372921.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | ENST00000372921.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | ENST00000372921.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | ENST00000372921.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | ENST00000372921.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | ENST00000372921.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | ENST00000372921.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | ENST00000394847.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | ENST00000394847.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | ENST00000394847.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | ENST00000394847.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | ENST00000394847.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | ENST00000394847.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | ENST00000463788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | ENST00000463788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | ENST00000463788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | ENST00000463788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | ENST00000463788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | ENST00000492380.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | ENST00000492380.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | ENST00000492380.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | ENST00000492380.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | ENST00000492380.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | ENST00000492380.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | ENST00000492380.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | ENST00000492380.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | ENST00000492380.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | NM_001156.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | NM_001156.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | NM_001156.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | NM_001156.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | NM_001156.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | NM_001156.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | NM_001156.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | NM_001156.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | NM_001156.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | NM_001156.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | NM_001156.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | NM_001156.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | NM_001156.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | NM_001156.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | NM_001156.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | NM_001156.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | NM_001156.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | NM_001156.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | NM_001156.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | NM_004034.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | NM_004034.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | NM_004034.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | NM_004034.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | NM_004034.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | NM_004034.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | NM_004034.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | NM_004034.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | NM_004034.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | NM_004034.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | NM_004034.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | NM_004034.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | NM_004034.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | NM_004034.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | NM_004034.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | NM_004034.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | NM_004034.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | NM_004034.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | NM_004034.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011020.hg.1 | NM_004034.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000175038.hg.1 | ENST00000372919.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000175038.hg.1 | NM_004034.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000175038.hg.1 | ENST00000372919.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000175038.hg.1 | NM_004034.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000175038.hg.1 | TC1000011020.hg.1 | Clariom D TC |

ANXA7
Genomic Map