| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01053325.hg.1 |
TC01003409.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01053325.hg.1 |
ENST00000326245.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01053325.hg.1 |
NM_017625.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01053326.hg.1 |
TC01003409.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01053326.hg.1 |
ENST00000326245.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01053326.hg.1 |
NM_017625.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016147.hg.1 |
ENST00000326245.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016147.hg.1 |
ENST00000326245.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016147.hg.1 |
ENST00000326245.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016147.hg.1 |
ENST00000326245.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016147.hg.1 |
ENST00000326245.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016147.hg.1 |
ENST00000326245.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016147.hg.1 |
ENST00000326245.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016147.hg.1 |
ENST00000326245.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016147.hg.1 |
ENST00000326245.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016147.hg.1 |
ENST00000464077.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016147.hg.1 |
ENST00000464077.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016147.hg.1 |
ENST00000464077.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016147.hg.1 |
ENST00000487531.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016147.hg.1 |
ENST00000487531.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016147.hg.1 |
ENST00000487531.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016147.hg.1 |
ENST00000487531.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016147.hg.1 |
NM_017625.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016147.hg.1 |
NM_017625.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016147.hg.1 |
NM_017625.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016147.hg.1 |
NM_017625.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016147.hg.1 |
NM_017625.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016147.hg.1 |
NM_017625.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016147.hg.1 |
NM_017625.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016147.hg.1 |
NM_017625.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016147.hg.1 |
NM_017625.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016147.hg.1 |
ENST00000326245.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016147.hg.1 |
ENST00000326245.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016147.hg.1 |
ENST00000326245.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016147.hg.1 |
ENST00000326245.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016147.hg.1 |
ENST00000326245.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016147.hg.1 |
ENST00000326245.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016147.hg.1 |
ENST00000326245.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016147.hg.1 |
ENST00000326245.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016147.hg.1 |
ENST00000326245.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016147.hg.1 |
ENST00000464077.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016147.hg.1 |
ENST00000464077.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016147.hg.1 |
ENST00000464077.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016147.hg.1 |
ENST00000487531.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016147.hg.1 |
ENST00000487531.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016147.hg.1 |
ENST00000487531.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016147.hg.1 |
ENST00000487531.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016147.hg.1 |
NM_017625.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016147.hg.1 |
NM_017625.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016147.hg.1 |
NM_017625.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016147.hg.1 |
NM_017625.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016147.hg.1 |
NM_017625.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016147.hg.1 |
NM_017625.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016147.hg.1 |
NM_017625.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016147.hg.1 |
NM_017625.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016147.hg.1 |
NM_017625.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100222791.hg.1 |
ENST00000326245.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100222791.hg.1 |
NM_017625.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100222791.hg.1 |
ENST00000326245.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100222791.hg.1 |
NM_017625.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100222791.hg.1 |
TC0100016147.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100222793.hg.1 |
ENST00000326245.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100222793.hg.1 |
NM_017625.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100222793.hg.1 |
ENST00000326245.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100222793.hg.1 |
NM_017625.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100222793.hg.1 |
TC0100016147.hg.1 |
Clariom D TC |