Assay Details
| Gene Symbol: SLC24A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020689.3 | NP_065740.2 | 8 - 9 | 914 | 122 | |
| GenBank mRNA | AF169257.2 | - | 8 - 9 | 660 | 122 | |
| AF288087.1 | - | 8 - 9 | 914 | 122 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20000130.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
ion transport
potassium ion transport
cellular calcium ion homeostasis
sodium ion transmembrane transport
calcium ion transmembrane transport
calcium channel activity
calcium ion binding
calcium, potassium:sodium antiporter activity
symporter activity
potassium ion binding
sodium ion binding
ion transport
potassium ion transport
cellular calcium ion homeostasis
sodium ion transmembrane transport
calcium ion transmembrane transport
calcium channel activity
calcium ion binding
calcium, potassium:sodium antiporter activity
symporter activity
potassium ion binding
sodium ion binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2000006852.hg.1 | ENST00000328041.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006852.hg.1 | ENST00000328041.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006852.hg.1 | ENST00000328041.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006852.hg.1 | ENST00000328041.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006852.hg.1 | ENST00000328041.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006852.hg.1 | ENST00000328041.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006852.hg.1 | ENST00000328041.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006852.hg.1 | ENST00000328041.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006852.hg.1 | ENST00000328041.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006852.hg.1 | ENST00000328041.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006852.hg.1 | ENST00000328041.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006852.hg.1 | ENST00000328041.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006852.hg.1 | ENST00000328041.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006852.hg.1 | ENST00000328041.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006852.hg.1 | ENST00000328041.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006852.hg.1 | ENST00000328041.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006852.hg.1 | ENST00000328041.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006852.hg.1 | ENST00000328041.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006852.hg.1 | ENST00000328041.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006852.hg.1 | ENST00000328041.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006852.hg.1 | ENST00000613834.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006852.hg.1 | ENST00000613834.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006852.hg.1 | ENST00000613834.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006852.hg.1 | ENST00000613834.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006852.hg.1 | ENST00000613834.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006852.hg.1 | ENST00000613834.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006852.hg.1 | ENST00000613834.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006852.hg.1 | ENST00000613834.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006852.hg.1 | ENST00000613834.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006852.hg.1 | ENST00000613834.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006852.hg.1 | ENST00000613834.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006852.hg.1 | ENST00000613834.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006852.hg.1 | ENST00000613834.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006852.hg.1 | ENST00000613834.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006852.hg.1 | ENST00000613834.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006852.hg.1 | ENST00000613834.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006852.hg.1 | ENST00000613834.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006852.hg.1 | ENST00000613834.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006852.hg.1 | NM_020689.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006852.hg.1 | NM_020689.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006852.hg.1 | NM_020689.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006852.hg.1 | NM_020689.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006852.hg.1 | NM_020689.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006852.hg.1 | NM_020689.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006852.hg.1 | NM_020689.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006852.hg.1 | NM_020689.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006852.hg.1 | NM_020689.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006852.hg.1 | NM_020689.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006852.hg.1 | NM_020689.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006852.hg.1 | NM_020689.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006852.hg.1 | NM_020689.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006852.hg.1 | NM_020689.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006852.hg.1 | NM_020689.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006852.hg.1 | NM_020689.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006852.hg.1 | NM_020689.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006852.hg.1 | NM_020689.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006852.hg.1 | NM_020689.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20001671.hg.1 | ENST00000328041.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20001671.hg.1 | NM_020689.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20001671.hg.1 | TC20000130.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR2000146888.hg.1 | ENST00000328041.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000146888.hg.1 | ENST00000613834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000146888.hg.1 | NM_020689.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000146888.hg.1 | TC2000006852.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000146888.hg.1 | ENST00000328041.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000146888.hg.1 | ENST00000613834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000146888.hg.1 | NM_020689.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006852.hg.1 | ENST00000328041.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006852.hg.1 | ENST00000328041.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006852.hg.1 | ENST00000328041.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006852.hg.1 | ENST00000328041.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006852.hg.1 | ENST00000328041.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006852.hg.1 | ENST00000328041.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006852.hg.1 | ENST00000328041.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006852.hg.1 | ENST00000328041.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006852.hg.1 | ENST00000328041.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006852.hg.1 | ENST00000328041.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006852.hg.1 | ENST00000328041.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006852.hg.1 | ENST00000328041.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006852.hg.1 | ENST00000328041.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006852.hg.1 | ENST00000328041.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006852.hg.1 | ENST00000328041.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006852.hg.1 | ENST00000328041.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006852.hg.1 | ENST00000328041.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006852.hg.1 | ENST00000328041.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006852.hg.1 | ENST00000328041.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006852.hg.1 | ENST00000328041.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006852.hg.1 | ENST00000613834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006852.hg.1 | ENST00000613834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006852.hg.1 | ENST00000613834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006852.hg.1 | ENST00000613834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006852.hg.1 | ENST00000613834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006852.hg.1 | ENST00000613834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006852.hg.1 | ENST00000613834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006852.hg.1 | ENST00000613834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006852.hg.1 | ENST00000613834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006852.hg.1 | ENST00000613834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006852.hg.1 | ENST00000613834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006852.hg.1 | ENST00000613834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006852.hg.1 | ENST00000613834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006852.hg.1 | ENST00000613834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006852.hg.1 | ENST00000613834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006852.hg.1 | ENST00000613834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006852.hg.1 | ENST00000613834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006852.hg.1 | ENST00000613834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006852.hg.1 | NM_020689.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006852.hg.1 | NM_020689.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006852.hg.1 | NM_020689.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006852.hg.1 | NM_020689.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006852.hg.1 | NM_020689.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006852.hg.1 | NM_020689.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006852.hg.1 | NM_020689.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006852.hg.1 | NM_020689.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006852.hg.1 | NM_020689.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006852.hg.1 | NM_020689.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006852.hg.1 | NM_020689.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006852.hg.1 | NM_020689.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006852.hg.1 | NM_020689.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006852.hg.1 | NM_020689.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006852.hg.1 | NM_020689.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006852.hg.1 | NM_020689.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006852.hg.1 | NM_020689.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006852.hg.1 | NM_020689.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006852.hg.1 | NM_020689.3 | Refseq |

SLC24A3
Genomic Map