Assay Details
| Gene Symbol: FGF17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003867.3 | NP_003858.1 | 2 - 3 | 246 | 83 | |
| GenBank mRNA | AB009249.1 | - | 2 - 3 | 82 | 83 | |
| BC069475.1 | - | 2 - 3 | 82 | 83 | ||
| BC105131.1 | - | 2 - 3 | 120 | 8144134 | 83 | |
| BC113489.1 | - | 2 - 3 | 120 | 8322541 | 83 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08000163.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
signal transduction
cell-cell signaling
nervous system development
positive regulation of cell proliferation
fibroblast growth factor receptor signaling pathway
regulation of phosphatidylinositol 3-kinase signaling
peptidyl-tyrosine phosphorylation
phosphatidylinositol-3-phosphate biosynthetic process
positive regulation of GTPase activity
phosphatidylinositol phosphorylation
phosphatidylinositol-mediated signaling
protein tyrosine kinase activity
Ras guanyl-nucleotide exchange factor activity
type 1 fibroblast growth factor receptor binding
type 2 fibroblast growth factor receptor binding
growth factor activity
1-phosphatidylinositol-3-kinase activity
phosphatidylinositol-4,5-bisphosphate 3-kinase activity
MAPK cascade
signal transduction
cell-cell signaling
nervous system development
positive regulation of cell proliferation
fibroblast growth factor receptor signaling pathway
regulation of phosphatidylinositol 3-kinase signaling
peptidyl-tyrosine phosphorylation
phosphatidylinositol-3-phosphate biosynthetic process
positive regulation of GTPase activity
phosphatidylinositol phosphorylation
phosphatidylinositol-mediated signaling
protein tyrosine kinase activity
Ras guanyl-nucleotide exchange factor activity
type 1 fibroblast growth factor receptor binding
type 2 fibroblast growth factor receptor binding
growth factor activity
1-phosphatidylinositol-3-kinase activity
phosphatidylinositol-4,5-bisphosphate 3-kinase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR08001714.hg.1 | TC08000163.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08001714.hg.1 | ENST00000359441.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08001714.hg.1 | ENST00000518533.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08001714.hg.1 | NM_003867.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006968.hg.1 | ENST00000359441.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006968.hg.1 | ENST00000359441.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006968.hg.1 | ENST00000359441.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006968.hg.1 | ENST00000359441.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006968.hg.1 | ENST00000359441.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006968.hg.1 | ENST00000359441.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006968.hg.1 | ENST00000359441.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006968.hg.1 | ENST00000359441.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006968.hg.1 | ENST00000359441.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006968.hg.1 | ENST00000359441.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006968.hg.1 | ENST00000359441.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006968.hg.1 | ENST00000359441.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006968.hg.1 | ENST00000518533.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006968.hg.1 | ENST00000518533.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006968.hg.1 | ENST00000518533.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006968.hg.1 | ENST00000518533.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006968.hg.1 | ENST00000518533.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006968.hg.1 | ENST00000518533.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006968.hg.1 | ENST00000518533.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006968.hg.1 | ENST00000518533.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006968.hg.1 | ENST00000518533.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006968.hg.1 | ENST00000518533.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006968.hg.1 | ENST00000518533.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006968.hg.1 | ENST00000518533.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006968.hg.1 | ENST00000521709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006968.hg.1 | ENST00000521709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006968.hg.1 | ENST00000521709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006968.hg.1 | ENST00000521709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006968.hg.1 | ENST00000524314.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006968.hg.1 | ENST00000524314.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006968.hg.1 | ENST00000524314.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006968.hg.1 | ENST00000524314.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006968.hg.1 | ENST00000524314.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006968.hg.1 | ENST00000524314.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006968.hg.1 | ENST00000524314.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006968.hg.1 | ENST00000524314.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006968.hg.1 | ENST00000524314.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006968.hg.1 | NM_001304478.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006968.hg.1 | NM_001304478.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006968.hg.1 | NM_001304478.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006968.hg.1 | NM_001304478.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006968.hg.1 | NM_001304478.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006968.hg.1 | NM_001304478.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006968.hg.1 | NM_001304478.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006968.hg.1 | NM_001304478.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006968.hg.1 | NM_001304478.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006968.hg.1 | NM_001304478.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006968.hg.1 | NM_003867.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006968.hg.1 | NM_003867.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006968.hg.1 | NM_003867.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006968.hg.1 | NM_003867.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006968.hg.1 | NM_003867.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006968.hg.1 | NM_003867.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006968.hg.1 | NM_003867.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006968.hg.1 | NM_003867.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006968.hg.1 | NM_003867.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006968.hg.1 | NM_003867.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006968.hg.1 | NM_003867.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800147257.hg.1 | ENST00000359441.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800147257.hg.1 | ENST00000518533.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800147257.hg.1 | ENST00000521709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800147257.hg.1 | ENST00000524314.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800147257.hg.1 | NM_001304478.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800147257.hg.1 | NM_003867.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800147257.hg.1 | ENST00000359441.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800147257.hg.1 | ENST00000518533.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800147257.hg.1 | ENST00000521709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800147257.hg.1 | ENST00000524314.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800147257.hg.1 | NM_001304478.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800147257.hg.1 | NM_003867.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800147257.hg.1 | TC0800006968.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0800006968.hg.1 | ENST00000359441.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006968.hg.1 | ENST00000359441.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006968.hg.1 | ENST00000359441.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006968.hg.1 | ENST00000359441.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006968.hg.1 | ENST00000359441.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006968.hg.1 | ENST00000359441.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006968.hg.1 | ENST00000359441.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006968.hg.1 | ENST00000359441.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006968.hg.1 | ENST00000359441.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006968.hg.1 | ENST00000359441.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006968.hg.1 | ENST00000359441.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006968.hg.1 | ENST00000359441.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006968.hg.1 | ENST00000518533.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006968.hg.1 | ENST00000518533.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006968.hg.1 | ENST00000518533.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006968.hg.1 | ENST00000518533.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006968.hg.1 | ENST00000518533.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006968.hg.1 | ENST00000518533.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006968.hg.1 | ENST00000518533.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006968.hg.1 | ENST00000518533.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006968.hg.1 | ENST00000518533.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006968.hg.1 | ENST00000518533.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006968.hg.1 | ENST00000518533.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006968.hg.1 | ENST00000518533.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006968.hg.1 | ENST00000521709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006968.hg.1 | ENST00000521709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006968.hg.1 | ENST00000521709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006968.hg.1 | ENST00000521709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006968.hg.1 | ENST00000524314.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006968.hg.1 | ENST00000524314.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006968.hg.1 | ENST00000524314.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006968.hg.1 | ENST00000524314.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006968.hg.1 | ENST00000524314.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006968.hg.1 | ENST00000524314.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006968.hg.1 | ENST00000524314.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006968.hg.1 | ENST00000524314.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006968.hg.1 | ENST00000524314.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006968.hg.1 | NM_001304478.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006968.hg.1 | NM_001304478.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006968.hg.1 | NM_001304478.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006968.hg.1 | NM_001304478.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006968.hg.1 | NM_001304478.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006968.hg.1 | NM_001304478.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006968.hg.1 | NM_001304478.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006968.hg.1 | NM_001304478.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006968.hg.1 | NM_001304478.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006968.hg.1 | NM_001304478.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006968.hg.1 | NM_003867.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006968.hg.1 | NM_003867.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006968.hg.1 | NM_003867.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006968.hg.1 | NM_003867.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006968.hg.1 | NM_003867.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006968.hg.1 | NM_003867.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006968.hg.1 | NM_003867.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006968.hg.1 | NM_003867.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006968.hg.1 | NM_003867.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006968.hg.1 | NM_003867.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006968.hg.1 | NM_003867.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800147256.hg.1 | ENST00000359441.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800147256.hg.1 | ENST00000524314.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800147256.hg.1 | NM_003867.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800147256.hg.1 | ENST00000359441.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800147256.hg.1 | ENST00000524314.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800147256.hg.1 | NM_003867.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800147256.hg.1 | TC0800006968.hg.1 | Clariom D TC |

FGF17
Genomic Map