Assay Details
| Gene Symbol: OPN5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181744.3 | NP_859528.1 | 2 - 3 | 276 | 100 | |
| XM_017010410.1 | XP_016865899.1 | 1 - 2 | 402 | 100 | ||
| XM_017010411.1 | XP_016865900.1 | 1 - 2 | 384 | 100 | ||
| XM_017010416.1 | XP_016865905.1 | 2 - 3 | 278 | 100 | ||
| GenBank mRNA | AY377391.1 | - | 2 - 3 | 253 | 100 | |
| BC126194.1 | - | 2 - 3 | 276 | 8991910 | 100 | |
| BC126198.1 | - | 2 - 3 | 276 | 8991914 | 100 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06000637.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | ENST00000244799.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | ENST00000244799.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | ENST00000244799.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | ENST00000244799.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | ENST00000244799.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | ENST00000244799.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | ENST00000244799.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | ENST00000244799.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | ENST00000244799.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | ENST00000244799.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | ENST00000244799.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | ENST00000244799.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | ENST00000244799.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | ENST00000371211.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | ENST00000371211.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | ENST00000371211.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | ENST00000371211.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | ENST00000371211.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | ENST00000371211.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | ENST00000371211.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | ENST00000371211.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | ENST00000371211.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | ENST00000371211.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | ENST00000371211.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | ENST00000371211.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | ENST00000371211.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | ENST00000393699.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | ENST00000393699.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | ENST00000393699.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | ENST00000393699.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | ENST00000393699.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | ENST00000393699.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | ENST00000393699.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | ENST00000393699.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | ENST00000393699.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | ENST00000393699.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | ENST00000393699.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | ENST00000489301.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | ENST00000489301.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | ENST00000489301.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | ENST00000489301.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | ENST00000489301.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | ENST00000489301.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | ENST00000489301.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | ENST00000489301.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | ENST00000489301.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | ENST00000489301.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | ENST00000489301.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | ENST00000489301.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | ENST00000489301.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | ENST00000510695.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | ENST00000510695.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | ENST00000510695.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | ENST00000510695.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | NM_181744.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | NM_181744.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | NM_181744.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | NM_181744.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | NM_181744.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | NM_181744.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | NM_181744.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | NM_181744.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | NM_181744.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | NM_181744.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | NM_181744.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | NM_181744.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | NM_181744.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | NM_181744.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | NR_033806.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | NR_033806.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | NR_033806.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | NR_033806.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | NR_033806.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | NR_033806.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | NR_033806.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | NR_033806.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | NR_033806.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | NR_033806.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | NR_033806.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | NR_033806.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008190.hg.1 | NR_033806.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06007758.hg.1 | TC06000637.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06007758.hg.1 | ENST00000244799.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06007758.hg.1 | ENST00000371211.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06007758.hg.1 | ENST00000393699.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06007758.hg.1 | ENST00000489301.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06007758.hg.1 | NM_181744.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06007758.hg.1 | NR_033806.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600157319.hg.1 | ENST00000244799.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600157319.hg.1 | ENST00000371211.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600157319.hg.1 | ENST00000393699.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600157319.hg.1 | ENST00000489301.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600157319.hg.1 | NM_181744.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600157319.hg.1 | NR_033806.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600157319.hg.1 | ENST00000244799.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600157319.hg.1 | ENST00000371211.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600157319.hg.1 | ENST00000393699.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600157319.hg.1 | ENST00000489301.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600157319.hg.1 | NM_181744.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600157319.hg.1 | NR_033806.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600157319.hg.1 | TC0600008190.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | ENST00000244799.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | ENST00000244799.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | ENST00000244799.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | ENST00000244799.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | ENST00000244799.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | ENST00000244799.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | ENST00000244799.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | ENST00000244799.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | ENST00000244799.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | ENST00000244799.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | ENST00000244799.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | ENST00000244799.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | ENST00000244799.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | ENST00000371211.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | ENST00000371211.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | ENST00000371211.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | ENST00000371211.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | ENST00000371211.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | ENST00000371211.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | ENST00000371211.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | ENST00000371211.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | ENST00000371211.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | ENST00000371211.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | ENST00000371211.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | ENST00000371211.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | ENST00000371211.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | ENST00000393699.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | ENST00000393699.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | ENST00000393699.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | ENST00000393699.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | ENST00000393699.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | ENST00000393699.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | ENST00000393699.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | ENST00000393699.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | ENST00000393699.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | ENST00000393699.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | ENST00000393699.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | ENST00000489301.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | ENST00000489301.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | ENST00000489301.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | ENST00000489301.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | ENST00000489301.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | ENST00000489301.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | ENST00000489301.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | ENST00000489301.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | ENST00000489301.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | ENST00000489301.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | ENST00000489301.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | ENST00000489301.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | ENST00000489301.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | ENST00000510695.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | ENST00000510695.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | ENST00000510695.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | ENST00000510695.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | NM_181744.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | NM_181744.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | NM_181744.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | NM_181744.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | NM_181744.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | NM_181744.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | NM_181744.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | NM_181744.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | NM_181744.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | NM_181744.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | NM_181744.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | NM_181744.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | NM_181744.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | NM_181744.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | NR_033806.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | NR_033806.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | NR_033806.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | NR_033806.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | NR_033806.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | NR_033806.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | NR_033806.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | NR_033806.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | NR_033806.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | NR_033806.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | NR_033806.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | NR_033806.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008190.hg.1 | NR_033806.1 | Refseq |

OPN5
Genomic Map