| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01039279.hg.1 |
TC01002525.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01039279.hg.1 |
ENST00000373044.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01039279.hg.1 |
NM_024640.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01039281.hg.1 |
TC01002525.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01039281.hg.1 |
ENST00000373044.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01039281.hg.1 |
NM_024640.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01039278.hg.1 |
TC01002525.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01039278.hg.1 |
ENST00000373044.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01039278.hg.1 |
NM_024640.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013756.hg.1 |
ENST00000373044.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013756.hg.1 |
ENST00000373044.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013756.hg.1 |
ENST00000373044.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013756.hg.1 |
ENST00000373044.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013756.hg.1 |
ENST00000373044.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013756.hg.1 |
ENST00000373044.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013756.hg.1 |
NM_024640.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013756.hg.1 |
NM_024640.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013756.hg.1 |
NM_024640.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013756.hg.1 |
NM_024640.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013756.hg.1 |
NM_024640.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013756.hg.1 |
NM_024640.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01039280.hg.1 |
TC01002525.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01039280.hg.1 |
ENST00000373044.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01039280.hg.1 |
NM_024640.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100202681.hg.1 |
ENST00000373044.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100202681.hg.1 |
NM_024640.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100202681.hg.1 |
ENST00000373044.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100202681.hg.1 |
NM_024640.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100202681.hg.1 |
TC0100013756.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100202678.hg.1 |
ENST00000373044.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100202678.hg.1 |
NM_024640.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100202678.hg.1 |
ENST00000373044.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100202678.hg.1 |
NM_024640.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100202678.hg.1 |
TC0100013756.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100202680.hg.1 |
ENST00000373044.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100202680.hg.1 |
NM_024640.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100202680.hg.1 |
ENST00000373044.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100202680.hg.1 |
NM_024640.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100202680.hg.1 |
TC0100013756.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100202679.hg.1 |
ENST00000373044.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100202679.hg.1 |
NM_024640.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100202679.hg.1 |
ENST00000373044.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100202679.hg.1 |
NM_024640.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100202679.hg.1 |
TC0100013756.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100013756.hg.1 |
ENST00000373044.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013756.hg.1 |
ENST00000373044.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013756.hg.1 |
ENST00000373044.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013756.hg.1 |
ENST00000373044.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013756.hg.1 |
ENST00000373044.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013756.hg.1 |
ENST00000373044.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013756.hg.1 |
NM_024640.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013756.hg.1 |
NM_024640.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013756.hg.1 |
NM_024640.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013756.hg.1 |
NM_024640.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013756.hg.1 |
NM_024640.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013756.hg.1 |
NM_024640.3 |
Refseq |