Assay Details
| Gene Symbol: ACRV1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001612.5 | NP_001603.1 | 2 | 248 | ||
| NM_020069.4 | NP_064454.1 | 2 | 248 | |||
| NM_020107.4 | NP_064492.1 | 2 - 3 | 239 | 83 | ||
| GenBank mRNA | AB451476.1 | - | 2 | 248 | ||
| AK223335.1 | - | 2 | 248 | |||
| BC014588.2 | - | 2 | 4825974 | 248 | ||
| BI826178.1 | - | 2 - 3 | 172 | 83 | ||
| M82967.1 | - | 2 | 248 | |||
| M82968.1 | - | 2 | 248 | |||
More Information
Set Membership: |
|

ACRV1
Genomic Map