Assay Details
| Gene Symbol: EIF2B4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034116.1 | NP_001029288.1 | 3 - 4 | 350 | 66 | |
| NM_001318965.1 | NP_001305894.1 | 2 - 3 | 309 | 66 | ||
| NM_001318966.1 | NP_001305895.1 | 3 - 4 | 400 | 66 | ||
| NM_001318968.1 | NP_001305897.1 | 2 - 3 | 309 | 66 | ||
| NM_001318969.1 | NP_001305898.1 | 3 - 4 | 350 | 66 | ||
| GenBank mRNA | AF112207.1 | - | 3 - 4 | 226 | 66 | |
| AF218009.1 | - | 3 - 4 | 371 | 66 | ||
| BC001870.1 | - | 3 - 4 | 208 | 3534267 | 66 | |
| BC091502.1 | - | 3 - 4 | 223 | 30377783 | 66 | |
| BG717490.1 | - | 2 - 3 | 288 | 66 | ||
| BI255289.1 | - | 2 - 3 | 268 | 66 | ||
| JQ991007.1 | - | 3 - 4 | 350 | 66 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001318968.1, NM_001318969.1 transcripts.
Set Membership: |
|

EIF2B4
Genomic Map