Assay Details
| Gene Symbol: UGCG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003358.2 | NP_003349.1 | 8 - 9 | 1461 | 91 | |
| XM_005252186.4 | XP_005252243.1 | 9 - 10 | 1484 | 91 | ||
| GenBank mRNA | AK314847.1 | - | 8 - 9 | 1066 | 91 | |
| BC038711.1 | - | 8 - 9 | 1462 | 5295561 | 91 | |
| D50840.1 | - | 8 - 9 | 1301 | 91 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09000560.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000474306.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000474306.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000474306.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000489355.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000489355.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000489355.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000490110.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000490110.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000490110.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000490110.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000490110.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000490110.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000495085.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000495085.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000495085.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000495085.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000495085.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000495085.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000495085.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000495085.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | NM_003358.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | NM_003358.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | NM_003358.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | NM_003358.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | NM_003358.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | NM_003358.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | NM_003358.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | NM_003358.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | NM_003358.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | NM_003358.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | NM_003358.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | NM_003358.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | NM_003358.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | NM_003358.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | NM_003358.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09006260.hg.1 | TC09000560.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09006260.hg.1 | ENST00000374279.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09006260.hg.1 | NM_003358.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900156439.hg.1 | ENST00000374279.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900156439.hg.1 | NM_003358.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900156439.hg.1 | ENST00000374279.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900156439.hg.1 | NM_003358.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900156439.hg.1 | TC0900008441.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900156441.hg.1 | ENST00000374279.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900156441.hg.1 | NM_003358.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900156441.hg.1 | ENST00000374279.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900156441.hg.1 | NM_003358.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900156441.hg.1 | TC0900008441.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900156440.hg.1 | ENST00000374279.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900156440.hg.1 | NM_003358.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900156440.hg.1 | ENST00000374279.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900156440.hg.1 | NM_003358.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900156440.hg.1 | TC0900008441.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000474306.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000474306.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000474306.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000489355.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000489355.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000489355.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000490110.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000490110.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000490110.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000490110.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000490110.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000490110.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000495085.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000495085.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000495085.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000495085.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000495085.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000495085.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000495085.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000495085.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | NM_003358.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | NM_003358.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | NM_003358.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | NM_003358.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | NM_003358.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | NM_003358.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | NM_003358.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | NM_003358.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | NM_003358.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | NM_003358.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | NM_003358.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | NM_003358.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | NM_003358.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | NM_003358.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | NM_003358.2 | Refseq |

UGCG
Genomic Map