Assay Details
| Gene Symbol: NCBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002486.4 | NP_002477.1 | 13 - 14 | 1756 | 136 | |
| XM_011518715.2 | XP_011517017.1 | 13 - 14 | 1755 | 136 | ||
| XM_017014735.1 | XP_016870224.1 | 13 - 14 | 1594 | 136 | ||
| XM_017014736.1 | XP_016870225.1 | 13 - 14 | 1307 | 136 | ||
| XM_017014737.1 | XP_016870226.1 | 13 - 14 | 1295 | 136 | ||
| XM_017014738.1 | XP_016870227.1 | 12 - 13 | 1295 | 136 | ||
| GenBank mRNA | AB209233.1 | - | 13 - 14 | 1336 | 136 | |
| AK312807.1 | - | 13 - 14 | 1756 | 136 | ||
| BC001450.2 | - | 13 - 14 | 1360 | 3138757 | 136 | |
| D32002.1 | - | 13 - 14 | 1561 | 136 | ||
| X80030.1 | - | 13 - 14 | 1327 | 136 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09000492.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Molecular Function -
Gene Ontology Categories:
Function(s)
Process(es)
nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
mRNA splicing, via spliceosome
transcription from RNA polymerase II promoter
transcription elongation from RNA polymerase II promoter
termination of RNA polymerase II transcription
7-methylguanosine mRNA capping
RNA export from nucleus
mRNA export from nucleus
regulation of translational initiation
histone mRNA metabolic process
RNA splicing
fibroblast growth factor receptor signaling pathway
gene expression
positive regulation of cell growth
gene silencing by RNA
mRNA 3'-end processing
positive regulation of mRNA 3'-end processing
snRNA transcription from RNA polymerase II promoter
mRNA cis splicing, via spliceosome
nuclear export
pre-mRNA cleavage required for polyadenylation
regulation of mRNA polyadenylation
RNA cap binding
RNA binding
protein binding
poly(A) RNA binding
nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
mRNA splicing, via spliceosome
transcription from RNA polymerase II promoter
transcription elongation from RNA polymerase II promoter
termination of RNA polymerase II transcription
7-methylguanosine mRNA capping
RNA export from nucleus
mRNA export from nucleus
regulation of translational initiation
histone mRNA metabolic process
RNA splicing
fibroblast growth factor receptor signaling pathway
gene expression
positive regulation of cell growth
gene silencing by RNA
mRNA 3'-end processing
positive regulation of mRNA 3'-end processing
snRNA transcription from RNA polymerase II promoter
mRNA cis splicing, via spliceosome
nuclear export
pre-mRNA cleavage required for polyadenylation
regulation of mRNA polyadenylation
RNA cap binding
RNA binding
protein binding
poly(A) RNA binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | ENST00000375130.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | ENST00000375130.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | ENST00000375130.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | ENST00000375130.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | ENST00000375147.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | ENST00000375147.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | ENST00000375147.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | ENST00000375147.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | ENST00000375147.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | ENST00000375147.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | ENST00000375147.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | ENST00000375147.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | ENST00000375147.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | ENST00000375147.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | ENST00000375147.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | ENST00000375147.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | ENST00000375147.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | ENST00000375147.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | ENST00000375147.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | ENST00000375147.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | ENST00000375147.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | ENST00000375147.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | ENST00000375147.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | ENST00000375147.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | ENST00000375147.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | ENST00000375147.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | ENST00000375147.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | ENST00000375147.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | ENST00000375147.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | ENST00000375147.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | ENST00000375147.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | ENST00000375147.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | ENST00000375147.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | ENST00000375147.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | ENST00000375147.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | ENST00000375147.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | ENST00000375147.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | ENST00000478100.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | ENST00000478100.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | ENST00000478100.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | ENST00000478100.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | ENST00000478100.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | ENST00000478100.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | ENST00000478100.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | ENST00000478100.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | ENST00000478100.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | ENST00000478100.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | ENST00000491445.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | ENST00000491445.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | ENST00000629069.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | ENST00000629069.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | ENST00000629069.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | ENST00000629069.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | NM_002486.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | NM_002486.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | NM_002486.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | NM_002486.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | NM_002486.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | NM_002486.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | NM_002486.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | NM_002486.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | NM_002486.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | NM_002486.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | NM_002486.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | NM_002486.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | NM_002486.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | NM_002486.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | NM_002486.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | NM_002486.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | NM_002486.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | NM_002486.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | NM_002486.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | NM_002486.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | NM_002486.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | NM_002486.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | NM_002486.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | NM_002486.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | NM_002486.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | NM_002486.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | NM_002486.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | NM_002486.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | NM_002486.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | NM_002486.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | NM_002486.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | NM_002486.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | NM_002486.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008156.hg.1 | NM_002486.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09005320.hg.1 | TC09000492.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09005320.hg.1 | ENST00000375147.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09005320.hg.1 | NM_002486.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | ENST00000375130.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | ENST00000375130.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | ENST00000375130.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | ENST00000375130.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | ENST00000375147.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | ENST00000375147.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | ENST00000375147.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | ENST00000375147.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | ENST00000375147.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | ENST00000375147.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | ENST00000375147.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | ENST00000375147.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | ENST00000375147.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | ENST00000375147.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | ENST00000375147.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | ENST00000375147.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | ENST00000375147.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | ENST00000375147.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | ENST00000375147.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | ENST00000375147.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | ENST00000375147.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | ENST00000375147.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | ENST00000375147.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | ENST00000375147.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | ENST00000375147.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | ENST00000375147.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | ENST00000375147.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | ENST00000375147.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | ENST00000375147.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | ENST00000375147.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | ENST00000375147.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | ENST00000375147.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | ENST00000375147.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | ENST00000375147.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | ENST00000375147.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | ENST00000375147.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | ENST00000375147.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | ENST00000478100.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | ENST00000478100.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | ENST00000478100.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | ENST00000478100.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | ENST00000478100.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | ENST00000478100.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | ENST00000478100.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | ENST00000478100.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | ENST00000478100.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | ENST00000478100.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | ENST00000491445.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | ENST00000491445.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | ENST00000629069.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | ENST00000629069.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | ENST00000629069.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | ENST00000629069.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | NM_002486.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | NM_002486.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | NM_002486.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | NM_002486.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | NM_002486.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | NM_002486.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | NM_002486.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | NM_002486.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | NM_002486.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | NM_002486.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | NM_002486.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | NM_002486.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | NM_002486.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | NM_002486.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | NM_002486.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | NM_002486.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | NM_002486.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | NM_002486.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | NM_002486.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | NM_002486.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | NM_002486.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | NM_002486.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | NM_002486.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | NM_002486.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | NM_002486.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | NM_002486.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | NM_002486.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | NM_002486.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | NM_002486.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | NM_002486.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | NM_002486.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | NM_002486.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | NM_002486.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008156.hg.1 | NM_002486.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900154703.hg.1 | ENST00000375147.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900154703.hg.1 | NM_002486.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900154703.hg.1 | ENST00000375147.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900154703.hg.1 | NM_002486.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900154703.hg.1 | TC0900008156.hg.1 | Clariom D TC |

NCBP1
Genomic Map