Assay Details
| Gene Symbol: ADSS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126.3 | NP_001117.2 | 8 - 9 | 1112 | 100 | |
| XM_011544108.2 | XP_011542410.1 | 8 - 9 | 1784 | 100 | ||
| GenBank mRNA | AK303886.1 | - | 9 - 10 | 824 | 100 | |
| BC012356.1 | - | 8 - 9 | 897 | 4635347 | 100 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01004046.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
immune system process
AMP biosynthetic process
aspartate metabolic process
purine ribonucleoside monophosphate biosynthetic process
response to purine-containing compound
'de novo' AMP biosynthetic process
IMP metabolic process
response to ammonium ion
cellular response to electrical stimulus
magnesium ion binding
adenylosuccinate synthase activity
protein binding
GTP binding
phosphate ion binding
immune system process
AMP biosynthetic process
aspartate metabolic process
purine ribonucleoside monophosphate biosynthetic process
response to purine-containing compound
'de novo' AMP biosynthetic process
IMP metabolic process
response to ammonium ion
cellular response to electrical stimulus
magnesium ion binding
adenylosuccinate synthase activity
protein binding
GTP binding
phosphate ion binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01061083.hg.1 | TC01004046.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01061083.hg.1 | ENST00000366535.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01061083.hg.1 | ENST00000449326.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01061083.hg.1 | NM_001126.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018006.hg.1 | ENST00000366535.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018006.hg.1 | ENST00000366535.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018006.hg.1 | ENST00000366535.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018006.hg.1 | ENST00000366535.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018006.hg.1 | ENST00000366535.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018006.hg.1 | ENST00000366535.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018006.hg.1 | ENST00000366535.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018006.hg.1 | ENST00000366535.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018006.hg.1 | ENST00000366535.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018006.hg.1 | ENST00000366535.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018006.hg.1 | ENST00000366535.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018006.hg.1 | ENST00000366535.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018006.hg.1 | ENST00000366535.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018006.hg.1 | ENST00000366535.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018006.hg.1 | ENST00000366535.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018006.hg.1 | ENST00000366535.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018006.hg.1 | ENST00000366535.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018006.hg.1 | ENST00000366535.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018006.hg.1 | ENST00000366535.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018006.hg.1 | ENST00000366535.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018006.hg.1 | ENST00000417765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018006.hg.1 | ENST00000417765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018006.hg.1 | ENST00000417765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018006.hg.1 | ENST00000417765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018006.hg.1 | ENST00000462358.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018006.hg.1 | ENST00000462358.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018006.hg.1 | ENST00000462358.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018006.hg.1 | ENST00000462358.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018006.hg.1 | ENST00000462358.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018006.hg.1 | ENST00000468215.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018006.hg.1 | ENST00000468215.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018006.hg.1 | ENST00000468215.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018006.hg.1 | ENST00000468215.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018006.hg.1 | ENST00000468215.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018006.hg.1 | NM_001126.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018006.hg.1 | NM_001126.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018006.hg.1 | NM_001126.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018006.hg.1 | NM_001126.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018006.hg.1 | NM_001126.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018006.hg.1 | NM_001126.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018006.hg.1 | NM_001126.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018006.hg.1 | NM_001126.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018006.hg.1 | NM_001126.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018006.hg.1 | NM_001126.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018006.hg.1 | NM_001126.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018006.hg.1 | NM_001126.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018006.hg.1 | NM_001126.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018006.hg.1 | NM_001126.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018006.hg.1 | NM_001126.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018006.hg.1 | NM_001126.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018006.hg.1 | NM_001126.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018006.hg.1 | NM_001126.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018006.hg.1 | NM_001126.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018006.hg.1 | NM_001126.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100235616.hg.1 | ENST00000366535.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100235616.hg.1 | ENST00000468215.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100235616.hg.1 | NM_001126.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100235616.hg.1 | ENST00000366535.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100235616.hg.1 | ENST00000468215.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100235616.hg.1 | NM_001126.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100235616.hg.1 | TC0100018006.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100018006.hg.1 | ENST00000366535.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018006.hg.1 | ENST00000366535.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018006.hg.1 | ENST00000366535.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018006.hg.1 | ENST00000366535.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018006.hg.1 | ENST00000366535.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018006.hg.1 | ENST00000366535.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018006.hg.1 | ENST00000366535.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018006.hg.1 | ENST00000366535.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018006.hg.1 | ENST00000366535.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018006.hg.1 | ENST00000366535.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018006.hg.1 | ENST00000366535.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018006.hg.1 | ENST00000366535.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018006.hg.1 | ENST00000366535.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018006.hg.1 | ENST00000366535.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018006.hg.1 | ENST00000366535.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018006.hg.1 | ENST00000366535.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018006.hg.1 | ENST00000366535.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018006.hg.1 | ENST00000366535.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018006.hg.1 | ENST00000366535.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018006.hg.1 | ENST00000366535.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018006.hg.1 | ENST00000417765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018006.hg.1 | ENST00000417765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018006.hg.1 | ENST00000417765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018006.hg.1 | ENST00000417765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018006.hg.1 | ENST00000462358.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018006.hg.1 | ENST00000462358.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018006.hg.1 | ENST00000462358.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018006.hg.1 | ENST00000462358.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018006.hg.1 | ENST00000462358.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018006.hg.1 | ENST00000468215.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018006.hg.1 | ENST00000468215.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018006.hg.1 | ENST00000468215.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018006.hg.1 | ENST00000468215.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018006.hg.1 | ENST00000468215.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018006.hg.1 | NM_001126.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018006.hg.1 | NM_001126.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018006.hg.1 | NM_001126.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018006.hg.1 | NM_001126.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018006.hg.1 | NM_001126.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018006.hg.1 | NM_001126.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018006.hg.1 | NM_001126.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018006.hg.1 | NM_001126.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018006.hg.1 | NM_001126.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018006.hg.1 | NM_001126.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018006.hg.1 | NM_001126.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018006.hg.1 | NM_001126.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018006.hg.1 | NM_001126.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018006.hg.1 | NM_001126.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018006.hg.1 | NM_001126.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018006.hg.1 | NM_001126.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018006.hg.1 | NM_001126.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018006.hg.1 | NM_001126.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018006.hg.1 | NM_001126.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018006.hg.1 | NM_001126.3 | Refseq |

ADSS
Genomic Map