Assay Details
| Gene Symbol: ACRV1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001612.5 | NP_001603.1 | 2 | 288 | ||
| NM_020069.4 | NP_064454.1 | 2 | 288 | |||
| NM_020107.4 | NP_064492.1 | 2 - 3 | 123 | |||
| NM_020108.4 | NP_064493.1 | 2 - 3 | 239 | 78 | ||
| GenBank mRNA | AB451476.1 | - | 2 | 288 | ||
| AK223335.1 | - | 2 | 288 | |||
| BC014588.2 | - | 2 | 4825974 | 288 | ||
| BG720361.1 | - | 2 - 3 | 209 | 78 | ||
| BI826178.1 | - | 2 - 3 | 123 | |||
| M82967.1 | - | 2 | 288 | |||
| M82968.1 | - | 2 | 288 | |||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11002422.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | ENST00000315608.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | ENST00000315608.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | ENST00000315608.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | ENST00000315608.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | ENST00000315608.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | ENST00000315608.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | ENST00000315608.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | ENST00000315608.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | ENST00000315608.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | ENST00000315608.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | ENST00000527795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | ENST00000527795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | ENST00000527795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | ENST00000527795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | ENST00000527795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | ENST00000527795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | ENST00000527795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | ENST00000527795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | ENST00000527795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | ENST00000530048.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | ENST00000530048.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | ENST00000530048.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | ENST00000530048.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | ENST00000530048.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | ENST00000530048.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | ENST00000530048.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | ENST00000530048.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | ENST00000530048.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | ENST00000533904.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | ENST00000533904.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | ENST00000533904.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | ENST00000533904.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | ENST00000533904.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | ENST00000533904.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | ENST00000533904.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | ENST00000533904.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | ENST00000533904.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | ENST00000533904.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | ENST00000533904.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | ENST00000533904.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | ENST00000533904.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | ENST00000533904.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | NM_001612.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | NM_001612.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | NM_001612.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | NM_001612.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | NM_001612.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | NM_001612.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | NM_001612.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | NM_001612.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | NM_001612.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | NM_001612.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | NM_001612.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | NM_001612.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | NM_020069.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | NM_020069.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | NM_020069.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | NM_020069.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | NM_020069.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | NM_020069.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | NM_020069.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | NM_020069.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | NM_020069.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | NM_020069.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | NM_020069.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | NM_020107.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | NM_020107.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | NM_020107.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | NM_020107.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | NM_020107.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | NM_020107.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | NM_020107.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | NM_020107.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | NM_020107.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | NM_020107.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | NM_020107.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | NM_020108.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | NM_020108.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | NM_020108.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | NM_020108.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | NM_020108.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | NM_020108.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | NM_020108.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | NM_020108.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | NM_020108.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012712.hg.1 | NM_020108.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029042.hg.1 | ENST00000315608.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029042.hg.1 | ENST00000433875.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029042.hg.1 | ENST00000533904.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029042.hg.1 | NM_001612.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029042.hg.1 | NM_020069.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029042.hg.1 | TC11002422.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029041.hg.1 | ENST00000257382.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029041.hg.1 | ENST00000315608.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029041.hg.1 | ENST00000348856.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029041.hg.1 | ENST00000433875.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029041.hg.1 | ENST00000530048.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029041.hg.1 | ENST00000533904.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029041.hg.1 | NM_001612.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029041.hg.1 | NM_020069.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029041.hg.1 | NM_020107.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029041.hg.1 | TC11002422.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | ENST00000315608.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | ENST00000315608.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | ENST00000315608.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | ENST00000315608.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | ENST00000315608.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | ENST00000315608.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | ENST00000315608.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | ENST00000315608.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | ENST00000315608.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | ENST00000315608.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | ENST00000527795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | ENST00000527795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | ENST00000527795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | ENST00000527795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | ENST00000527795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | ENST00000527795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | ENST00000527795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | ENST00000527795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | ENST00000527795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | ENST00000530048.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | ENST00000530048.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | ENST00000530048.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | ENST00000530048.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | ENST00000530048.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | ENST00000530048.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | ENST00000530048.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | ENST00000530048.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | ENST00000530048.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | ENST00000533904.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | ENST00000533904.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | ENST00000533904.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | ENST00000533904.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | ENST00000533904.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | ENST00000533904.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | ENST00000533904.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | ENST00000533904.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | ENST00000533904.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | ENST00000533904.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | ENST00000533904.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | ENST00000533904.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | ENST00000533904.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | ENST00000533904.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | NM_001612.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | NM_001612.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | NM_001612.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | NM_001612.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | NM_001612.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | NM_001612.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | NM_001612.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | NM_001612.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | NM_001612.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | NM_001612.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | NM_001612.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | NM_001612.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | NM_020069.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | NM_020069.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | NM_020069.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | NM_020069.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | NM_020069.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | NM_020069.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | NM_020069.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | NM_020069.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | NM_020069.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | NM_020069.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | NM_020069.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | NM_020107.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | NM_020107.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | NM_020107.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | NM_020107.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | NM_020107.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | NM_020107.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | NM_020107.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | NM_020107.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | NM_020107.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | NM_020107.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | NM_020107.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | NM_020108.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | NM_020108.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | NM_020108.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | NM_020108.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | NM_020108.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | NM_020108.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | NM_020108.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | NM_020108.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | NM_020108.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012712.hg.1 | NM_020108.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100197752.hg.1 | ENST00000315608.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100197752.hg.1 | ENST00000530048.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100197752.hg.1 | ENST00000533904.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100197752.hg.1 | NM_001612.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100197752.hg.1 | NM_020069.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100197752.hg.1 | NM_020107.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100197752.hg.1 | ENST00000315608.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100197752.hg.1 | ENST00000530048.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100197752.hg.1 | ENST00000533904.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100197752.hg.1 | NM_001612.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100197752.hg.1 | NM_020069.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100197752.hg.1 | NM_020107.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100197752.hg.1 | TC1100012712.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100197753.hg.1 | ENST00000315608.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100197753.hg.1 | ENST00000533904.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100197753.hg.1 | NM_001612.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100197753.hg.1 | NM_020069.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100197753.hg.1 | ENST00000315608.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100197753.hg.1 | ENST00000533904.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100197753.hg.1 | NM_001612.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100197753.hg.1 | NM_020069.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100197753.hg.1 | TC1100012712.hg.1 | Clariom D TC |

ACRV1
Genomic Map