Assay Details
| Gene Symbol: TNS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022648.5 | NP_072174.3 | 20 - 21 | 3394 | 65 | |
| XM_017004804.1 | XP_016860293.1 | 19 - 20 | 3401 | 65 | ||
| GenBank mRNA | BC140942.1 | - | 20 - 21 | 3326 | 9056754 | 65 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

TNS1
Genomic Map