Assay Details
| Gene Symbol: TNS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022648.5 | NP_072174.3 | 21 - 22 | 3420 | 59 | |
| XM_017004804.1 | XP_016860293.1 | 20 - 21 | 3427 | 59 | ||
| GenBank mRNA | BC140942.1 | - | 21 - 22 | 3352 | 9056754 | 59 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02002765.hg.1
Set Membership: |
|

TNS1
Genomic Map