Assay Details
| Gene Symbol: MAGOH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK297895.1 | - | 2 - 3 | 227 | 140 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100014227.hg.1 | ENST00000371466.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014227.hg.1 | ENST00000371466.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014227.hg.1 | ENST00000371466.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014227.hg.1 | ENST00000371466.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014227.hg.1 | ENST00000371466.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014227.hg.1 | ENST00000371466.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014227.hg.1 | ENST00000371470.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014227.hg.1 | ENST00000371470.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014227.hg.1 | ENST00000371470.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014227.hg.1 | ENST00000371470.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014227.hg.1 | ENST00000371470.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014227.hg.1 | ENST00000371470.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014227.hg.1 | ENST00000371470.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014227.hg.1 | ENST00000371470.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014227.hg.1 | ENST00000371470.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014227.hg.1 | ENST00000462941.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014227.hg.1 | ENST00000462941.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014227.hg.1 | ENST00000462941.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014227.hg.1 | ENST00000462941.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014227.hg.1 | ENST00000462941.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014227.hg.1 | ENST00000495868.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014227.hg.1 | ENST00000495868.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014227.hg.1 | ENST00000495868.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014227.hg.1 | ENST00000495868.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014227.hg.1 | ENST00000495868.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014227.hg.1 | NM_002370.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014227.hg.1 | NM_002370.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014227.hg.1 | NM_002370.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014227.hg.1 | NM_002370.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014227.hg.1 | NM_002370.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014227.hg.1 | NM_002370.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014227.hg.1 | NM_002370.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014227.hg.1 | NM_002370.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014227.hg.1 | NM_002370.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014227.hg.1 | ENST00000371466.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014227.hg.1 | ENST00000371466.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014227.hg.1 | ENST00000371466.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014227.hg.1 | ENST00000371466.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014227.hg.1 | ENST00000371466.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014227.hg.1 | ENST00000371466.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014227.hg.1 | ENST00000371470.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014227.hg.1 | ENST00000371470.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014227.hg.1 | ENST00000371470.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014227.hg.1 | ENST00000371470.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014227.hg.1 | ENST00000371470.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014227.hg.1 | ENST00000371470.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014227.hg.1 | ENST00000371470.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014227.hg.1 | ENST00000371470.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014227.hg.1 | ENST00000371470.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014227.hg.1 | ENST00000462941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014227.hg.1 | ENST00000462941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014227.hg.1 | ENST00000462941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014227.hg.1 | ENST00000462941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014227.hg.1 | ENST00000462941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014227.hg.1 | ENST00000495868.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014227.hg.1 | ENST00000495868.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014227.hg.1 | ENST00000495868.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014227.hg.1 | ENST00000495868.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014227.hg.1 | ENST00000495868.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014227.hg.1 | NM_002370.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014227.hg.1 | NM_002370.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014227.hg.1 | NM_002370.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014227.hg.1 | NM_002370.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014227.hg.1 | NM_002370.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014227.hg.1 | NM_002370.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014227.hg.1 | NM_002370.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014227.hg.1 | NM_002370.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014227.hg.1 | NM_002370.3 | Refseq |

MAGOH
Genomic Map