Assay Details
| Gene Symbol: UGGT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020120.3 | NP_064505.1 | 21 - 22 | 2439 | 117 | |
| NR_027671.2 | 21 - 22 | 2592 | 117 | |||
| XM_006712634.3 | XP_006712697.1 | 21 - 22 | 2413 | 117 | ||
| XM_006712636.3 | XP_006712699.1 | 21 - 22 | 2402 | 117 | ||
| XM_017004507.1 | XP_016859996.1 | 22 - 23 | 2498 | 117 | ||
| XM_017004508.1 | XP_016859997.1 | 22 - 23 | 2453 | 117 | ||
| XM_017004509.1 | XP_016859998.1 | 7 - 8 | 744 | 117 | ||
| XM_017004510.1 | XP_016859999.1 | 3 - 4 | 1123 | 117 | ||
| GenBank mRNA | AF227905.1 | - | 21 - 22 | 2299 | 117 | |
| AK023671.1 | - | 2 - 3 | 135 | 117 | ||
| AK074251.1 | - | 5 - 6 | 463 | 117 | ||
| AK293066.1 | - | 21 - 22 | 2592 | 117 | ||
| AK309101.1 | - | 19 - 20 | 2164 | 117 | ||
| BC041098.1 | - | 21 - 22 | 2589 | 5492334 | 117 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02000791.hg
Additional Information:
This assay is in the 5'UTR region of the AK023671.1,XM_017004510.1 transcripts.
Set Membership: |
|

UGGT1
Genomic Map