Assay Details
| Gene Symbol: MAP1LC3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK310224.1 | - | 2 - 3 | 208 | 108 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | ENST00000268607.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | ENST00000268607.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | ENST00000268607.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | ENST00000268607.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | ENST00000268607.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | ENST00000268607.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | ENST00000268607.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | ENST00000268607.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | ENST00000268607.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | ENST00000564638.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | ENST00000564638.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | ENST00000564638.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | ENST00000564844.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | ENST00000564844.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | ENST00000564844.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | ENST00000564844.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | ENST00000564844.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | ENST00000564844.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | ENST00000564844.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | ENST00000564844.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | ENST00000564844.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | ENST00000564844.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | ENST00000565788.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | ENST00000565788.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | ENST00000565788.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | ENST00000565788.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | ENST00000565788.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | ENST00000570189.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | ENST00000570189.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | ENST00000570189.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | ENST00000570189.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | ENST00000570189.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | ENST00000570189.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | ENST00000570189.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | NM_022818.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | NM_022818.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | NM_022818.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | NM_022818.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | NM_022818.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | NM_022818.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | NM_022818.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | NM_022818.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | ENST00000268607.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | ENST00000268607.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | ENST00000268607.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | ENST00000268607.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | ENST00000268607.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | ENST00000268607.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | ENST00000268607.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | ENST00000268607.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | ENST00000268607.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | ENST00000564638.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | ENST00000564638.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | ENST00000564638.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | ENST00000564844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | ENST00000564844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | ENST00000564844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | ENST00000564844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | ENST00000564844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | ENST00000564844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | ENST00000564844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | ENST00000564844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | ENST00000564844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | ENST00000564844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | ENST00000565788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | ENST00000565788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | ENST00000565788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | ENST00000565788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | ENST00000565788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | ENST00000570189.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | ENST00000570189.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | ENST00000570189.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | ENST00000570189.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | ENST00000570189.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | ENST00000570189.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | ENST00000570189.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | NM_022818.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | NM_022818.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | NM_022818.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | NM_022818.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | NM_022818.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | NM_022818.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | NM_022818.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | NM_022818.4 | Refseq |

MAP1LC3B
Genomic Map