Assay Details
| Gene Symbol: MAP1LC3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK310224.1 | - | 3 - 4 | 298 | 84 | |
| BC045759.1 | - | 3 - 4 | 672 | 4828857 | 84 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC045759.1 transcripts.
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | ENST00000268607.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | ENST00000268607.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | ENST00000268607.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | ENST00000268607.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | ENST00000268607.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | ENST00000268607.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | ENST00000268607.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | ENST00000268607.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | ENST00000268607.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | ENST00000564638.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | ENST00000564638.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | ENST00000564638.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | ENST00000564844.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | ENST00000564844.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | ENST00000564844.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | ENST00000564844.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | ENST00000564844.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | ENST00000564844.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | ENST00000564844.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | ENST00000564844.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | ENST00000564844.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | ENST00000564844.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | ENST00000565788.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | ENST00000565788.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | ENST00000565788.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | ENST00000565788.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | ENST00000565788.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | ENST00000570189.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | ENST00000570189.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | ENST00000570189.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | ENST00000570189.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | ENST00000570189.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | ENST00000570189.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | ENST00000570189.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | NM_022818.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | NM_022818.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | NM_022818.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | NM_022818.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | NM_022818.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | NM_022818.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | NM_022818.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011442.hg.1 | NM_022818.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008976.hg.1 | ENST00000548774.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008976.hg.1 | ENST00000548774.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009659.hg.1 | ENST00000417653.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009659.hg.1 | ENST00000417653.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009659.hg.1 | ENST00000438342.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009659.hg.1 | ENST00000438342.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009659.hg.1 | ENST00000438342.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009659.hg.1 | ENST00000438342.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009659.hg.1 | ENST00000438342.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191702.hg.1 | ENST00000268607.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191702.hg.1 | ENST00000564844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191702.hg.1 | ENST00000565788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191702.hg.1 | ENST00000570189.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191702.hg.1 | NM_022818.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600191702.hg.1 | ENST00000268607.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191702.hg.1 | ENST00000564844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191702.hg.1 | ENST00000565788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191702.hg.1 | ENST00000570189.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191702.hg.1 | NM_022818.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600191702.hg.1 | TC1600011442.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | ENST00000268607.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | ENST00000268607.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | ENST00000268607.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | ENST00000268607.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | ENST00000268607.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | ENST00000268607.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | ENST00000268607.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | ENST00000268607.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | ENST00000268607.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | ENST00000564638.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | ENST00000564638.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | ENST00000564638.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | ENST00000564844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | ENST00000564844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | ENST00000564844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | ENST00000564844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | ENST00000564844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | ENST00000564844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | ENST00000564844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | ENST00000564844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | ENST00000564844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | ENST00000564844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | ENST00000565788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | ENST00000565788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | ENST00000565788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | ENST00000565788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | ENST00000565788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | ENST00000570189.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | ENST00000570189.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | ENST00000570189.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | ENST00000570189.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | ENST00000570189.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | ENST00000570189.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | ENST00000570189.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | NM_022818.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | NM_022818.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | NM_022818.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | NM_022818.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | NM_022818.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | NM_022818.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | NM_022818.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011442.hg.1 | NM_022818.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600191698.hg.1 | ENST00000564844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191698.hg.1 | ENST00000564844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191698.hg.1 | TC1600011442.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200008976.hg.1 | ENST00000548774.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008976.hg.1 | ENST00000548774.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009659.hg.1 | ENST00000417653.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009659.hg.1 | ENST00000417653.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009659.hg.1 | ENST00000438342.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009659.hg.1 | ENST00000438342.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009659.hg.1 | ENST00000438342.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009659.hg.1 | ENST00000438342.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009659.hg.1 | ENST00000438342.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191704.hg.1 | ENST00000268607.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191704.hg.1 | ENST00000564844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191704.hg.1 | ENST00000570189.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191704.hg.1 | NM_022818.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600191704.hg.1 | ENST00000268607.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191704.hg.1 | ENST00000564844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191704.hg.1 | ENST00000570189.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191704.hg.1 | NM_022818.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600191704.hg.1 | TC1600011442.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600191705.hg.1 | ENST00000268607.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191705.hg.1 | ENST00000564844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191705.hg.1 | ENST00000570189.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191705.hg.1 | NM_022818.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600191705.hg.1 | ENST00000268607.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191705.hg.1 | ENST00000564844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191705.hg.1 | ENST00000570189.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191705.hg.1 | NM_022818.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600191705.hg.1 | TC1600011442.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600191701.hg.1 | ENST00000564844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191701.hg.1 | ENST00000570189.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191701.hg.1 | ENST00000564844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191701.hg.1 | ENST00000570189.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191701.hg.1 | TC1600011442.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600191699.hg.1 | ENST00000268607.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191699.hg.1 | ENST00000564638.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191699.hg.1 | ENST00000564844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191699.hg.1 | ENST00000565788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191699.hg.1 | ENST00000570189.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191699.hg.1 | NM_022818.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600191699.hg.1 | ENST00000268607.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191699.hg.1 | ENST00000564638.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191699.hg.1 | ENST00000564844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191699.hg.1 | ENST00000565788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191699.hg.1 | ENST00000570189.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191699.hg.1 | NM_022818.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600191699.hg.1 | TC1600011442.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 106979 | - | - |

MAP1LC3B
Genomic Map