Assay Details
| Gene Symbol: FAM228A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040710.2 | NP_001035800.1 | 1 - 2 | 138 | 131 | |
| GenBank mRNA | HY022970.1 | - | 1 - 2 | 139 | 131 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02005059.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_001040710.2 transcripts.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02001507.hg.1 | TC02005059.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001507.hg.1 | ENST00000295150.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001507.hg.1 | NM_001040710.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001510.hg.1 | TC02005059.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001510.hg.1 | ENST00000295150.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001510.hg.1 | NM_001040710.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001505.hg.1 | TC02005059.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001505.hg.1 | ENST00000295150.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001505.hg.1 | NM_001040710.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016415.hg.1 | ENST00000295150.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016415.hg.1 | ENST00000295150.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016415.hg.1 | ENST00000295150.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016415.hg.1 | ENST00000295150.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016415.hg.1 | ENST00000295150.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016415.hg.1 | ENST00000295150.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016415.hg.1 | ENST00000295150.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016415.hg.1 | ENST00000295150.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016415.hg.1 | ENST00000295150.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016415.hg.1 | ENST00000295150.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016415.hg.1 | ENST00000415196.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016415.hg.1 | ENST00000415196.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016415.hg.1 | ENST00000415196.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016415.hg.1 | ENST00000415196.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016415.hg.1 | ENST00000432434.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016415.hg.1 | ENST00000432434.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016415.hg.1 | ENST00000432434.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016415.hg.1 | ENST00000432434.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016415.hg.1 | ENST00000432434.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016415.hg.1 | ENST00000432434.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016415.hg.1 | ENST00000432434.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016415.hg.1 | ENST00000456591.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016415.hg.1 | ENST00000456591.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016415.hg.1 | ENST00000456591.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016415.hg.1 | ENST00000456591.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016415.hg.1 | ENST00000456591.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016415.hg.1 | ENST00000456591.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016415.hg.1 | ENST00000456591.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016415.hg.1 | ENST00000456591.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016415.hg.1 | ENST00000456591.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016415.hg.1 | ENST00000456591.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016415.hg.1 | NM_001040710.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016415.hg.1 | NM_001040710.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016415.hg.1 | NM_001040710.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016415.hg.1 | NM_001040710.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016415.hg.1 | NM_001040710.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016415.hg.1 | NM_001040710.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016415.hg.1 | NM_001040710.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016415.hg.1 | NM_001040710.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016415.hg.1 | NM_001040710.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016415.hg.1 | NM_001040710.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016415.hg.1 | NM_001040710.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016415.hg.1 | NM_001040710.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001503.hg.1 | TC02005059.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001503.hg.1 | ENST00000295150.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001503.hg.1 | NM_001040710.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200221192.hg.1 | ENST00000295150.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221192.hg.1 | ENST00000456591.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221192.hg.1 | NM_001040710.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200221192.hg.1 | ENST00000295150.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221192.hg.1 | ENST00000456591.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221192.hg.1 | NM_001040710.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200221192.hg.1 | TC0200016415.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200221199.hg.1 | ENST00000295150.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221199.hg.1 | ENST00000432434.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221199.hg.1 | ENST00000456591.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221199.hg.1 | NM_001040710.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200221199.hg.1 | ENST00000295150.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221199.hg.1 | ENST00000432434.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221199.hg.1 | ENST00000456591.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221199.hg.1 | NM_001040710.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200221199.hg.1 | TC0200016415.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200221190.hg.1 | NM_001040710.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200221190.hg.1 | NM_001040710.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200221190.hg.1 | TC0200016415.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200221195.hg.1 | ENST00000295150.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221195.hg.1 | ENST00000432434.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221195.hg.1 | ENST00000456591.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221195.hg.1 | NM_001040710.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200221195.hg.1 | ENST00000295150.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221195.hg.1 | ENST00000432434.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221195.hg.1 | ENST00000456591.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221195.hg.1 | NM_001040710.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200221195.hg.1 | TC0200016415.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200221196.hg.1 | ENST00000295150.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221196.hg.1 | ENST00000432434.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221196.hg.1 | ENST00000456591.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221196.hg.1 | NM_001040710.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200221196.hg.1 | ENST00000295150.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221196.hg.1 | ENST00000432434.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221196.hg.1 | ENST00000456591.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221196.hg.1 | NM_001040710.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200221196.hg.1 | TC0200016415.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200221191.hg.1 | ENST00000456591.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221191.hg.1 | NM_001040710.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200221191.hg.1 | ENST00000456591.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221191.hg.1 | NM_001040710.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200221191.hg.1 | TC0200016415.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200016415.hg.1 | ENST00000295150.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016415.hg.1 | ENST00000295150.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016415.hg.1 | ENST00000295150.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016415.hg.1 | ENST00000295150.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016415.hg.1 | ENST00000295150.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016415.hg.1 | ENST00000295150.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016415.hg.1 | ENST00000295150.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016415.hg.1 | ENST00000295150.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016415.hg.1 | ENST00000295150.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016415.hg.1 | ENST00000295150.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016415.hg.1 | ENST00000415196.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016415.hg.1 | ENST00000415196.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016415.hg.1 | ENST00000415196.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016415.hg.1 | ENST00000415196.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016415.hg.1 | ENST00000432434.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016415.hg.1 | ENST00000432434.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016415.hg.1 | ENST00000432434.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016415.hg.1 | ENST00000432434.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016415.hg.1 | ENST00000432434.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016415.hg.1 | ENST00000432434.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016415.hg.1 | ENST00000432434.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016415.hg.1 | ENST00000456591.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016415.hg.1 | ENST00000456591.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016415.hg.1 | ENST00000456591.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016415.hg.1 | ENST00000456591.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016415.hg.1 | ENST00000456591.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016415.hg.1 | ENST00000456591.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016415.hg.1 | ENST00000456591.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016415.hg.1 | ENST00000456591.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016415.hg.1 | ENST00000456591.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016415.hg.1 | ENST00000456591.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016415.hg.1 | NM_001040710.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016415.hg.1 | NM_001040710.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016415.hg.1 | NM_001040710.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016415.hg.1 | NM_001040710.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016415.hg.1 | NM_001040710.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016415.hg.1 | NM_001040710.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016415.hg.1 | NM_001040710.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016415.hg.1 | NM_001040710.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016415.hg.1 | NM_001040710.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016415.hg.1 | NM_001040710.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016415.hg.1 | NM_001040710.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016415.hg.1 | NM_001040710.2 | Refseq |

FAM228A
Genomic Map