Assay Details
| Gene Symbol: C1orf145 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_073154.1 | 1 - 2 | 239 | 87 | ||
| GenBank mRNA | AK056556.1 | - | 1 - 2 | 239 | 87 | |
| Gene Symbol: OBSCN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BU180714.1 | - | 229 | 87 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
IBA57 homolog, iron-sulfur cluster assembly |
IBA57 |
C1orf69, MMDS3, SPG74 |
-14.6kb | Anti-sense | Chr. 1 - 228165808 - 228182257 on NCBI Build GRCh38 |
| Downstream | MAGE-like protein 2 | LOC101927401 | 147.2kb | Sense | Chr. 1 - 228360893 - 228369935 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

C1orf145
Genomic Map