Assay Details
| Gene Symbol: EXOC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK074086.1 | - | 5 - 6 | 1647 | 71 | |
More Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000315013.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000315013.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000315013.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000315013.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000315013.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000315013.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000315013.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000315013.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000315013.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000315013.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000315013.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000315013.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000315013.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000315013.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000315013.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000505947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000505947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000505947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000505947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000508022.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000508022.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000508022.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000508022.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000510028.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000510028.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000510028.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000510028.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000510441.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000510441.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000510441.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000510441.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000510441.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000511015.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000511015.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000511015.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000511015.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000511015.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000512944.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000512944.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000512944.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000512944.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000512944.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000512944.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000512944.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000512944.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000512944.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000512944.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000512944.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000512944.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000512944.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000512944.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000512944.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000512944.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000512944.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | NM_007277.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | NM_007277.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | NM_007277.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | NM_007277.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | NM_007277.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | NM_007277.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | NM_007277.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | NM_007277.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | NM_007277.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | NM_007277.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | NM_007277.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | NM_007277.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | NM_007277.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | NM_007277.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | NM_007277.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | NM_007277.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000315013.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000315013.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000315013.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000315013.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000315013.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000315013.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000315013.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000315013.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000315013.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000315013.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000315013.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000315013.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000315013.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000315013.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000315013.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000505947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000505947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000505947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000505947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000508022.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000508022.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000508022.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000508022.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000510028.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000510028.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000510028.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000510028.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000510441.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000510441.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000510441.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000510441.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000510441.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000511015.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000511015.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000511015.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000511015.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000511015.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000512944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000512944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000512944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000512944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000512944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000512944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000512944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000512944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000512944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000512944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000512944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000512944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000512944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000512944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000512944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000512944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000512944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | NM_007277.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | NM_007277.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | NM_007277.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | NM_007277.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | NM_007277.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | NM_007277.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | NM_007277.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | NM_007277.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | NM_007277.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | NM_007277.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | NM_007277.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | NM_007277.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | NM_007277.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | NM_007277.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | NM_007277.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | NM_007277.4 | Refseq |

EXOC3
Genomic Map