Assay Details
| Gene Symbol: PIGZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025163.3 | NP_079439.2 | 1 - 2 | 188 | 63 | |
| XM_017007242.1 | XP_016862731.1 | 1 - 2 | 728 | 63 | ||
| GenBank mRNA | AK022830.1 | - | 1 - 2 | 150 | 63 | |
| BC018804.1 | - | 1 - 2 | 69 | 63 | ||
| BC044640.1 | - | 1 - 2 | 70 | 4871581 | 63 | |
| BG760692.1 | - | 1 - 2 | 52 | 63 | ||
| DA736224.1 | - | 1 - 2 | 188 | 63 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03002171.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK022830.1,BC044640.1,NM_025163.3,XM_017007242.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03038355.hg.1 | TC03002171.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03038355.hg.1 | ENST00000412723.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03038355.hg.1 | ENST00000443835.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03038355.hg.1 | NM_025163.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013728.hg.1 | ENST00000238138.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013728.hg.1 | ENST00000238138.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013728.hg.1 | ENST00000238138.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013728.hg.1 | ENST00000238138.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013728.hg.1 | ENST00000238138.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013728.hg.1 | ENST00000238138.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013728.hg.1 | ENST00000238138.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013728.hg.1 | ENST00000412723.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013728.hg.1 | ENST00000412723.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013728.hg.1 | ENST00000412723.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013728.hg.1 | ENST00000412723.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013728.hg.1 | ENST00000412723.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013728.hg.1 | ENST00000412723.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013728.hg.1 | ENST00000412723.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013728.hg.1 | ENST00000412723.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013728.hg.1 | ENST00000412723.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013728.hg.1 | ENST00000412723.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013728.hg.1 | ENST00000413127.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013728.hg.1 | ENST00000413127.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013728.hg.1 | ENST00000413127.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013728.hg.1 | ENST00000413127.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013728.hg.1 | ENST00000413127.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013728.hg.1 | ENST00000443835.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013728.hg.1 | ENST00000443835.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013728.hg.1 | ENST00000443835.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013728.hg.1 | ENST00000443835.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013728.hg.1 | ENST00000443835.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013728.hg.1 | ENST00000443835.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013728.hg.1 | NM_025163.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013728.hg.1 | NM_025163.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013728.hg.1 | NM_025163.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013728.hg.1 | NM_025163.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013728.hg.1 | NM_025163.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013728.hg.1 | NM_025163.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013728.hg.1 | NM_025163.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013728.hg.1 | NM_025163.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013728.hg.1 | NM_025163.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013728.hg.1 | NM_025163.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013728.hg.1 | NM_025163.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03038353.hg.1 | TC03002171.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03038353.hg.1 | ENST00000412723.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03038353.hg.1 | ENST00000443835.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03038353.hg.1 | NM_025163.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03038354.hg.1 | TC03002171.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03038354.hg.1 | ENST00000412723.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03038354.hg.1 | ENST00000443835.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03038354.hg.1 | NM_025163.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03038356.hg.1 | TC03002171.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03038356.hg.1 | ENST00000412723.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03038356.hg.1 | ENST00000443835.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03038356.hg.1 | NM_025163.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300202595.hg.1 | ENST00000238138.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300202595.hg.1 | ENST00000412723.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300202595.hg.1 | ENST00000413127.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300202595.hg.1 | ENST00000443835.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300202595.hg.1 | NM_025163.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300202595.hg.1 | ENST00000238138.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300202595.hg.1 | ENST00000412723.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300202595.hg.1 | ENST00000413127.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300202595.hg.1 | ENST00000443835.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300202595.hg.1 | NM_025163.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300202595.hg.1 | TC0300013728.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300202597.hg.1 | ENST00000238138.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300202597.hg.1 | ENST00000412723.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300202597.hg.1 | ENST00000443835.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300202597.hg.1 | NM_025163.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300202597.hg.1 | ENST00000238138.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300202597.hg.1 | ENST00000412723.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300202597.hg.1 | ENST00000443835.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300202597.hg.1 | NM_025163.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300202597.hg.1 | TC0300013728.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300202598.hg.1 | ENST00000238138.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300202598.hg.1 | ENST00000412723.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300202598.hg.1 | ENST00000443835.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300202598.hg.1 | NM_025163.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300202598.hg.1 | ENST00000238138.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300202598.hg.1 | ENST00000412723.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300202598.hg.1 | ENST00000443835.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300202598.hg.1 | NM_025163.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300202598.hg.1 | TC0300013728.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300202596.hg.1 | ENST00000238138.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300202596.hg.1 | ENST00000412723.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300202596.hg.1 | ENST00000443835.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300202596.hg.1 | NM_025163.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300202596.hg.1 | ENST00000238138.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300202596.hg.1 | ENST00000412723.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300202596.hg.1 | ENST00000443835.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300202596.hg.1 | NM_025163.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300202596.hg.1 | TC0300013728.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300202599.hg.1 | ENST00000238138.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300202599.hg.1 | ENST00000443835.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300202599.hg.1 | NM_025163.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300202599.hg.1 | ENST00000238138.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300202599.hg.1 | ENST00000443835.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300202599.hg.1 | NM_025163.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300202599.hg.1 | TC0300013728.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300013728.hg.1 | ENST00000238138.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013728.hg.1 | ENST00000238138.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013728.hg.1 | ENST00000238138.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013728.hg.1 | ENST00000238138.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013728.hg.1 | ENST00000238138.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013728.hg.1 | ENST00000238138.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013728.hg.1 | ENST00000238138.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013728.hg.1 | ENST00000412723.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013728.hg.1 | ENST00000412723.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013728.hg.1 | ENST00000412723.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013728.hg.1 | ENST00000412723.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013728.hg.1 | ENST00000412723.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013728.hg.1 | ENST00000412723.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013728.hg.1 | ENST00000412723.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013728.hg.1 | ENST00000412723.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013728.hg.1 | ENST00000412723.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013728.hg.1 | ENST00000412723.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013728.hg.1 | ENST00000413127.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013728.hg.1 | ENST00000413127.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013728.hg.1 | ENST00000413127.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013728.hg.1 | ENST00000413127.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013728.hg.1 | ENST00000413127.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013728.hg.1 | ENST00000443835.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013728.hg.1 | ENST00000443835.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013728.hg.1 | ENST00000443835.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013728.hg.1 | ENST00000443835.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013728.hg.1 | ENST00000443835.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013728.hg.1 | ENST00000443835.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013728.hg.1 | NM_025163.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013728.hg.1 | NM_025163.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013728.hg.1 | NM_025163.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013728.hg.1 | NM_025163.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013728.hg.1 | NM_025163.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013728.hg.1 | NM_025163.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013728.hg.1 | NM_025163.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013728.hg.1 | NM_025163.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013728.hg.1 | NM_025163.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013728.hg.1 | NM_025163.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013728.hg.1 | NM_025163.3 | Refseq |

PIGZ
Genomic Map