Assay Details
| Gene Symbol: MRPS5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001321996.1 | NP_001308925.1 | 1 - 2 | 296 | 76 | |
| NM_031902.4 | NP_114108.1 | 1 - 2 | 296 | 76 | ||
| NR_136152.1 | 1 - 2 | 296 | 76 | |||
| NR_136153.1 | 1 - 2 | 296 | 76 | |||
| GenBank mRNA | AB049940.1 | - | 1 - 2 | 177 | 76 | |
| AK058160.1 | - | 1 - 2 | 70 | 76 | ||
| AK295812.1 | - | 1 - 2 | 82 | 76 | ||
| BC014172.2 | - | 1 - 2 | 269 | 4561399 | 76 | |
| BG771419.1 | - | 1 - 2 | 283 | 76 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02004930.hg
Additional Information:
This assay is in the 5'UTR region of the NM_001321996.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02033755.hg.1 | TC02004930.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02033755.hg.1 | ENST00000272418.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02033755.hg.1 | NM_031902.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02033752.hg.1 | TC02004930.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02033752.hg.1 | ENST00000272418.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02033752.hg.1 | NM_031902.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02033753.hg.1 | TC02004930.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02033753.hg.1 | ENST00000272418.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02033753.hg.1 | NM_031902.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | ENST00000272418.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | ENST00000272418.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | ENST00000272418.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | ENST00000272418.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | ENST00000272418.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | ENST00000272418.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | ENST00000272418.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | ENST00000272418.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | ENST00000272418.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | ENST00000272418.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | ENST00000272418.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | ENST00000272418.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | ENST00000272418.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | ENST00000272418.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | ENST00000272418.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | ENST00000272418.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | ENST00000345084.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | ENST00000345084.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | ENST00000345084.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | ENST00000345084.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | ENST00000345084.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | ENST00000345084.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | ENST00000345084.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | ENST00000345084.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | ENST00000345084.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | ENST00000345084.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | ENST00000345084.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | ENST00000345084.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | ENST00000345084.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | ENST00000345084.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | ENST00000461916.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | ENST00000461916.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | ENST00000461916.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | ENST00000461916.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | ENST00000461916.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | ENST00000461916.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | ENST00000461916.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | ENST00000461916.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | ENST00000464458.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | ENST00000464458.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | ENST00000464458.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | ENST00000464458.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | ENST00000464458.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | ENST00000475895.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | ENST00000475895.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | ENST00000475895.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | ENST00000475895.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | ENST00000475895.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | ENST00000475895.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | ENST00000475895.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | ENST00000475895.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | ENST00000475895.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | ENST00000482568.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | ENST00000482568.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | ENST00000482568.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | ENST00000482568.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | ENST00000482568.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | ENST00000482568.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | ENST00000482568.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | ENST00000482821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | ENST00000482821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | ENST00000482821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | ENST00000482821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | ENST00000482821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | ENST00000482821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | ENST00000482821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | ENST00000482821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | NM_031902.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | NM_031902.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | NM_031902.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | NM_031902.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | NM_031902.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | NM_031902.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | NM_031902.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | NM_031902.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | NM_031902.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | NM_031902.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | NM_031902.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | NM_031902.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | NM_031902.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | NM_031902.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | NM_031902.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016706.hg.1 | NM_031902.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | ENST00000272418.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | ENST00000272418.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | ENST00000272418.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | ENST00000272418.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | ENST00000272418.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | ENST00000272418.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | ENST00000272418.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | ENST00000272418.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | ENST00000272418.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | ENST00000272418.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | ENST00000272418.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | ENST00000272418.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | ENST00000272418.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | ENST00000272418.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | ENST00000272418.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | ENST00000272418.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | ENST00000345084.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | ENST00000345084.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | ENST00000345084.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | ENST00000345084.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | ENST00000345084.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | ENST00000345084.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | ENST00000345084.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | ENST00000345084.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | ENST00000345084.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | ENST00000345084.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | ENST00000345084.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | ENST00000345084.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | ENST00000345084.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | ENST00000345084.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | ENST00000461916.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | ENST00000461916.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | ENST00000461916.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | ENST00000461916.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | ENST00000461916.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | ENST00000461916.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | ENST00000461916.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | ENST00000461916.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | ENST00000464458.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | ENST00000464458.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | ENST00000464458.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | ENST00000464458.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | ENST00000464458.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | ENST00000475895.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | ENST00000475895.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | ENST00000475895.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | ENST00000475895.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | ENST00000475895.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | ENST00000475895.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | ENST00000475895.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | ENST00000475895.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | ENST00000475895.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | ENST00000482568.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | ENST00000482568.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | ENST00000482568.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | ENST00000482568.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | ENST00000482568.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | ENST00000482568.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | ENST00000482568.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | ENST00000482821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | ENST00000482821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | ENST00000482821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | ENST00000482821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | ENST00000482821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | ENST00000482821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | ENST00000482821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | ENST00000482821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | NM_031902.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | NM_031902.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | NM_031902.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | NM_031902.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | NM_031902.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | NM_031902.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | NM_031902.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | NM_031902.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | NM_031902.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | NM_031902.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | NM_031902.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | NM_031902.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | NM_031902.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | NM_031902.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | NM_031902.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016706.hg.1 | NM_031902.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200227219.hg.1 | ENST00000272418.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200227219.hg.1 | ENST00000345084.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200227219.hg.1 | ENST00000475895.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200227219.hg.1 | NM_031902.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200227219.hg.1 | ENST00000272418.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200227219.hg.1 | ENST00000345084.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200227219.hg.1 | ENST00000475895.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200227219.hg.1 | NM_031902.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200227219.hg.1 | TC0200016706.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200227223.hg.1 | ENST00000272418.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200227223.hg.1 | NM_031902.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200227223.hg.1 | ENST00000272418.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200227223.hg.1 | NM_031902.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200227223.hg.1 | TC0200016706.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200227220.hg.1 | ENST00000272418.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200227220.hg.1 | ENST00000345084.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200227220.hg.1 | ENST00000475895.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200227220.hg.1 | NM_031902.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200227220.hg.1 | ENST00000272418.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200227220.hg.1 | ENST00000345084.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200227220.hg.1 | ENST00000475895.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200227220.hg.1 | NM_031902.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200227220.hg.1 | TC0200016706.hg.1 | Clariom D TC |

MRPS5
Genomic Map