Assay Details
| Gene Symbol: ORMDL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320801.1 | NP_001307730.1 | 6 | 2248 | 77 | |
| NM_001320802.1 | NP_001307731.1 | 4 | 1086 | 77 | ||
| NM_001320803.1 | NP_001307732.1 | 4 | 800 | 77 | ||
| NM_139280.3 | NP_644809.1 | 4 | 1091 | 77 | ||
| XM_017025345.1 | XP_016880834.1 | 4 | 944 | 77 | ||
| XM_017025346.1 | XP_016880835.1 | 4 | 857 | 77 | ||
| XM_017025347.1 | XP_016880836.1 | 4 | 1037 | 77 | ||
| XM_017025348.1 | XP_016880837.1 | 4 | 783 | 77 | ||
| GenBank mRNA | AF373101.1 | - | 4 | 827 | 77 | |
| AK074811.1 | - | 4 | 888 | 77 | ||
| AX775843.1 | - | 4 | 845 | 77 | ||
| BC017087.1 | - | 4 | 857 | 3850211 | 77 | |
| BC071833.1 | - | 4 | 836 | 6340437 | 77 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17002852.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | ENST00000304046.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | ENST00000304046.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | ENST00000304046.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | ENST00000304046.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | ENST00000304046.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | ENST00000304046.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | ENST00000304046.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | ENST00000304046.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | ENST00000304046.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | ENST00000304046.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | ENST00000304046.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | ENST00000304046.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | ENST00000304046.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | ENST00000304046.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | ENST00000394169.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | ENST00000394169.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | ENST00000394169.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | ENST00000394169.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | ENST00000394169.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | ENST00000394169.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | ENST00000394169.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | ENST00000394169.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | ENST00000394169.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | ENST00000394169.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | ENST00000394169.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | ENST00000394169.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | ENST00000394169.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | ENST00000579287.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | ENST00000579287.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | ENST00000579695.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | ENST00000579695.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | ENST00000579695.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | ENST00000579695.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | ENST00000579695.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | ENST00000579695.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | ENST00000579695.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | ENST00000579695.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | ENST00000579695.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | ENST00000579695.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | ENST00000579695.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | ENST00000579695.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | ENST00000579695.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | ENST00000579695.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | ENST00000581284.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | ENST00000581284.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | ENST00000581284.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | ENST00000581284.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | ENST00000581284.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | ENST00000582052.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | ENST00000582052.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | ENST00000584000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | ENST00000584000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | ENST00000584000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | ENST00000584000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | ENST00000584000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | ENST00000584220.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | ENST00000584220.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | ENST00000584220.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | ENST00000584220.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | NM_139280.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | NM_139280.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | NM_139280.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | NM_139280.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | NM_139280.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | NM_139280.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | NM_139280.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | NM_139280.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | NM_139280.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | NM_139280.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | NM_139280.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | NM_139280.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | NM_139280.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | NM_139280.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010592.hg.1 | NM_139280.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019232.hg.1 | TC17002852.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019232.hg.1 | ENST00000304046.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019232.hg.1 | NM_139280.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700183483.hg.1 | ENST00000304046.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700183483.hg.1 | ENST00000579695.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700183483.hg.1 | NM_139280.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700183483.hg.1 | ENST00000304046.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700183483.hg.1 | ENST00000579695.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700183483.hg.1 | NM_139280.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700183483.hg.1 | TC1700010592.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | ENST00000304046.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | ENST00000304046.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | ENST00000304046.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | ENST00000304046.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | ENST00000304046.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | ENST00000304046.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | ENST00000304046.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | ENST00000304046.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | ENST00000304046.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | ENST00000304046.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | ENST00000304046.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | ENST00000304046.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | ENST00000304046.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | ENST00000304046.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | ENST00000394169.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | ENST00000394169.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | ENST00000394169.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | ENST00000394169.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | ENST00000394169.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | ENST00000394169.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | ENST00000394169.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | ENST00000394169.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | ENST00000394169.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | ENST00000394169.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | ENST00000394169.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | ENST00000394169.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | ENST00000394169.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | ENST00000579287.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | ENST00000579287.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | ENST00000579695.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | ENST00000579695.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | ENST00000579695.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | ENST00000579695.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | ENST00000579695.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | ENST00000579695.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | ENST00000579695.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | ENST00000579695.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | ENST00000579695.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | ENST00000579695.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | ENST00000579695.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | ENST00000579695.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | ENST00000579695.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | ENST00000579695.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | ENST00000581284.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | ENST00000581284.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | ENST00000581284.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | ENST00000581284.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | ENST00000581284.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | ENST00000582052.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | ENST00000582052.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | ENST00000584000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | ENST00000584000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | ENST00000584000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | ENST00000584000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | ENST00000584000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | ENST00000584220.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | ENST00000584220.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | ENST00000584220.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | ENST00000584220.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | NM_139280.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | NM_139280.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | NM_139280.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | NM_139280.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | NM_139280.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | NM_139280.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | NM_139280.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | NM_139280.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | NM_139280.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | NM_139280.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | NM_139280.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | NM_139280.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | NM_139280.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | NM_139280.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010592.hg.1 | NM_139280.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700183484.hg.1 | ENST00000304046.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700183484.hg.1 | ENST00000579695.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700183484.hg.1 | ENST00000581284.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700183484.hg.1 | NM_139280.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700183484.hg.1 | ENST00000304046.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700183484.hg.1 | ENST00000579695.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700183484.hg.1 | ENST00000581284.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700183484.hg.1 | NM_139280.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700183484.hg.1 | TC1700010592.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700183486.hg.1 | ENST00000304046.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700183486.hg.1 | ENST00000394169.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700183486.hg.1 | ENST00000579695.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700183486.hg.1 | NM_139280.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700183486.hg.1 | ENST00000304046.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700183486.hg.1 | ENST00000394169.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700183486.hg.1 | ENST00000579695.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700183486.hg.1 | NM_139280.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700183486.hg.1 | TC1700010592.hg.1 | Clariom D TC |

ORMDL3
Genomic Map