Assay Details
| Gene Symbol: IGHD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK090461.1 | - | 6 - 7 | 1904 | 62 | |
| AK126280.1 | - | 7 - 8 | 1255 | 62 | ||
| AK128568.1 | - | 8 - 9 | 1271 | 62 | ||
| AK128580.1 | - | 7 - 8 | 1255 | 62 | ||
| AK128614.1 | - | 7 - 8 | 1312 | 62 | ||
| AK128625.1 | - | 7 - 8 | 1322 | 62 | ||
| AK128635.1 | - | 7 - 8 | 1333 | 62 | ||
| AK128658.1 | - | 6 - 7 | 1331 | 62 | ||
| BC021276.2 | - | 7 - 8 | 1281 | 62 | ||
| BC063384.1 | - | 7 - 8 | 1242 | 62 | ||
More Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P324838 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 25009 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 5130 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 41895 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P135384 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P324838 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P324838 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P135384 | - | - |
| Custom Human Agilent Array 11k, Chromosome 6, Expression | 2369 | - | - |
| Custom Human Agilent Array 11k, Chromosome 6, Expression | 58 | - | - |
| Custom Human Agilent Array 11k, Chromosome 6, Expression | 6891 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 7981627 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7981632 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7981626 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7981629 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7981660 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7981628 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7981631 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7981643 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7981633 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7981671 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7981725 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7981661 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7981724 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7981601 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 215621_PM_s_at | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 230877_PM_at | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 222285_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010449.hg.1 | ENST00000390556.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010449.hg.1 | ENST00000390556.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010449.hg.1 | ENST00000390556.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010449.hg.1 | ENST00000390556.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010449.hg.1 | ENST00000390556.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010449.hg.1 | ENST00000390556.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010449.hg.1 | ENST00000390556.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010449.hg.1 | ENST00000613640.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010449.hg.1 | ENST00000613640.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010449.hg.1 | ENST00000613640.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010449.hg.1 | ENST00000613640.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010449.hg.1 | ENST00000613640.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010449.hg.1 | ENST00000613640.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010706.hg.1 | ENST00000414005.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010706.hg.1 | ENST00000414005.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010706.hg.1 | ENST00000418566.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010706.hg.1 | ENST00000418566.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010706.hg.1 | ENST00000418566.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010706.hg.1 | ENST00000418566.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010706.hg.1 | ENST00000414005.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010706.hg.1 | ENST00000414005.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010706.hg.1 | ENST00000418566.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010706.hg.1 | ENST00000418566.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010706.hg.1 | ENST00000418566.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010706.hg.1 | ENST00000418566.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010449.hg.1 | ENST00000390556.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010449.hg.1 | ENST00000390556.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010449.hg.1 | ENST00000390556.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010449.hg.1 | ENST00000390556.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010449.hg.1 | ENST00000390556.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010449.hg.1 | ENST00000390556.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010449.hg.1 | ENST00000390556.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010449.hg.1 | ENST00000613640.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010449.hg.1 | ENST00000613640.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010449.hg.1 | ENST00000613640.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010449.hg.1 | ENST00000613640.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010449.hg.1 | ENST00000613640.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010449.hg.1 | ENST00000613640.1 | Ensembl |
| Human Genome U133 Plus 2.0 Array | 213674_PM_x_at | - | - |
| Human Genome U133 Plus 2.0 Array | 211637_x_at | - | - |
| Human Genome U133 Plus 2.0 Array | 211835_at | - | - |
| Human Genome U133 Plus 2.0 Array | 234419_PM_x_at | - | - |
| Human Genome U133 Plus 2.0 Array | 217217_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 230877_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 216510_x_at | - | - |
| Human Genome U133 Plus 2.0 Array | 213674_x_at | - | - |
| Human Genome U133 Plus 2.0 Array | 211632_at | - | - |
| Human Genome U133 Plus 2.0 Array | 217236_PM_x_at | - | - |
| Human Genome U133 Plus 2.0 Array | 215621_PM_s_at | - | - |
| Human Genome U133 Plus 2.0 Array | 217198_x_at | - | - |
| Human Genome U133 Plus 2.0 Array | 211638_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 211650_PM_x_at | - | - |
| Human Genome U133 Plus 2.0 Array | 230877_at | - | - |
| Human Genome U133 Plus 2.0 Array | 234419_x_at | - | - |
| Human Genome U133 Plus 2.0 Array | 216706_PM_x_at | - | - |
| Human Genome U133 Plus 2.0 Array | 211637_PM_x_at | - | - |
| Human Genome U133 Plus 2.0 Array | 216706_x_at | - | - |
| Human Genome U133 Plus 2.0 Array | 222285_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 216558_x_at | - | - |
| Human Genome U133 Plus 2.0 Array | 214916_PM_x_at | - | - |
| Human Genome U133 Plus 2.0 Array | 217198_PM_x_at | - | - |
| Human Genome U133 Plus 2.0 Array | 214973_x_at | - | - |
| Human Genome U133 Plus 2.0 Array | 215621_s_at | - | - |
| Human Genome U133 Plus 2.0 Array | 211638_at | - | - |
| Human Genome U133 Plus 2.0 Array | 211650_x_at | - | - |
| Human Genome U133 Plus 2.0 Array | 211835_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 217217_at | - | - |
| Human Genome U133 Plus 2.0 Array | 217236_x_at | - | - |
| Human Genome U133 Plus 2.0 Array | 211639_PM_x_at | - | - |
| Human Genome U133 Plus 2.0 Array | 222285_at | - | - |
| Human Genome U133 Plus 2.0 Array | 214916_x_at | - | - |
| Human Genome U133 Plus 2.0 Array | 214973_PM_x_at | - | - |
| Human Genome U133 Plus 2.0 Array | 211632_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 216558_PM_x_at | - | - |
| Human Genome U133 Plus 2.0 Array | 216510_PM_x_at | - | - |
| Human Genome U133 Plus 2.0 Array | 211639_x_at | - | - |
| Human Genome U133A 2.0 Array | 217217_PM_at | - | - |
| Human Genome U133A 2.0 Array | 211638_at | - | - |
| Human Genome U133A 2.0 Array | 211639_PM_x_at | - | - |
| Human Genome U133A 2.0 Array | 217217_at | - | - |
| Human Genome U133A 2.0 Array | 214916_PM_x_at | - | - |
| Human Genome U133A 2.0 Array | 217198_x_at | - | - |
| Human Genome U133A 2.0 Array | 211835_at | - | - |
| Human Genome U133A 2.0 Array | 211638_PM_at | - | - |
| Human Genome U133A 2.0 Array | 216706_x_at | - | - |
| Human Genome U133A 2.0 Array | 211632_PM_at | - | - |
| Human Genome U133A 2.0 Array | 211835_PM_at | - | - |
| Human Genome U133A 2.0 Array | 217236_x_at | - | - |
| Human Genome U133A 2.0 Array | 211632_at | - | - |
| Human Genome U133A 2.0 Array | 216558_PM_x_at | - | - |
| Human Genome U133A 2.0 Array | 216706_PM_x_at | - | - |
| Human Genome U133A 2.0 Array | 217236_PM_x_at | - | - |
| Human Genome U133A 2.0 Array | 214916_x_at | - | - |
| Human Genome U133A 2.0 Array | 217198_PM_x_at | - | - |
| Human Genome U133A 2.0 Array | 211637_PM_x_at | - | - |
| Human Genome U133A 2.0 Array | 216510_x_at | - | - |
| Human Genome U133A 2.0 Array | 216510_PM_x_at | - | - |
| Human Genome U133A 2.0 Array | 211637_x_at | - | - |
| Human Genome U133A 2.0 Array | 211650_x_at | - | - |
| Human Genome U133A 2.0 Array | 211639_x_at | - | - |
| Human Genome U133A 2.0 Array | 216558_x_at | - | - |
| Human Genome U133A 2.0 Array | 211650_PM_x_at | - | - |
| AffymetrixPathwork Diagnostics Pathchip | pw583306 | - | - |
| AffymetrixPathwork Diagnostics Pathchip | pw527083 | - | - |
| AffymetrixPathwork Diagnostics Pathchip | pw564316 | - | - |
| AffymetrixPathwork Diagnostics Pathchip | pw558589 | - | - |
| Human Genome U95 Set | 35607_PM_at | - | - |
| Human Genome U95 Set | 31317_r_at | - | - |
| Human Genome U95 Set | 31317_PM_r_at | - | - |
| Human Genome U95 Set | 35607_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 230877_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214973_x_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 230877_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214973_x_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 222285_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 213674_x_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 215621_s_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214973_x_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 213674_x_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 215621_s_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 230877_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 230877_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214973_x_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 230877_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 213674_x_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 222285_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 222285_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 213674_x_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214973_x_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 230877_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 213674_x_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214973_x_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 230877_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 213674_x_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 213674_x_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 222285_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 230877_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 215621_s_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 215621_s_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 222285_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 215621_s_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 213674_x_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 222285_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 230877_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 222285_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 215621_s_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 230877_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214973_x_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 215621_s_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 213674_x_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 222285_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 230877_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 213674_x_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214973_x_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 213674_x_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214973_x_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 222285_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 215621_s_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214973_x_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 222285_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 215621_s_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 215621_s_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 222285_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 215621_s_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214973_x_at8 | - | - |
| Human Genome U133 Set | 214916_x_at | - | - |
| Human Genome U133 Set | 216510_x_at | - | - |
| Human Genome U133 Set | 217198_x_at | - | - |
| Human Genome U133 Set | 216706_PM_x_at | - | - |
| Human Genome U133 Set | 217236_x_at | - | - |
| Human Genome U133 Set | 216558_PM_x_at | - | - |
| Human Genome U133 Set | 214916_PM_x_at | - | - |
| Human Genome U133 Set | 216510_PM_x_at | - | - |
| Human Genome U133 Set | 211639_x_at | - | - |
| Human Genome U133 Set | 211638_at | - | - |
| Human Genome U133 Set | 211639_PM_x_at | - | - |
| Human Genome U133 Set | 211632_at | - | - |
| Human Genome U133 Set | 217198_PM_x_at | - | - |
| Human Genome U133 Set | 217236_PM_x_at | - | - |
| Human Genome U133 Set | 211835_PM_at | - | - |
| Human Genome U133 Set | 211637_PM_x_at | - | - |
| Human Genome U133 Set | 217217_PM_at | - | - |
| Human Genome U133 Set | 211638_PM_at | - | - |
| Human Genome U133 Set | 216706_x_at | - | - |
| Human Genome U133 Set | 216558_x_at | - | - |
| Human Genome U133 Set | 211650_PM_x_at | - | - |
| Human Genome U133 Set | 211637_x_at | - | - |
| Human Genome U133 Set | 217217_at | - | - |
| Human Genome U133 Set | 211632_PM_at | - | - |
| Human Genome U133 Set | 211650_x_at | - | - |
| Human Genome U133 Set | 211835_at | - | - |
| Human Genome U133 Set | 234419_x_at | - | - |
| Human Genome U133 Set | 234419_PM_x_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina human-6 v2.0 expression beadchip | ILMN_1827611 | - | - |
| Illumina Sentrix Human-6 v2 Expression BeadChip | 7570187 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_114118 | - | - |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1876766 | - | - |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1876766 | - | - |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1827611 | - | - |

IGHD
Genomic Map