Assay Details
| Gene Symbol: SF3A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320830.1 | NP_001307759.1 | 8 - 9 | 812 | 61 | |
| NM_006802.3 | NP_006793.1 | 10 - 11 | 971 | 61 | ||
| GenBank mRNA | AK128438.1 | - | 10 - 11 | 1780 | 61 | |
| AK222554.1 | - | 10 - 11 | 847 | 61 | ||
| AK291181.1 | - | 10 - 11 | 852 | 61 | ||
| AK292116.1 | - | 10 - 11 | 882 | 61 | ||
| AK301424.1 | - | 8 - 9 | 689 | 61 | ||
| BC002395.2 | - | 10 - 11 | 864 | 2821350 | 61 | |
| BC011523.1 | - | 10 - 11 | 928 | 3867442 | 61 | |
| U08815.1 | - | 10 - 11 | 834 | 61 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01002529.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01039369.hg.1 | TC01002529.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01039369.hg.1 | ENST00000373019.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01039369.hg.1 | ENST00000448721.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01039369.hg.1 | ENST00000489537.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01039369.hg.1 | NM_006802.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | ENST00000373019.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | ENST00000373019.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | ENST00000373019.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | ENST00000373019.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | ENST00000373019.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | ENST00000373019.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | ENST00000373019.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | ENST00000373019.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | ENST00000373019.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | ENST00000373019.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | ENST00000373019.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | ENST00000373019.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | ENST00000373019.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | ENST00000373019.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | ENST00000373019.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | ENST00000373019.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | ENST00000373019.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | ENST00000373019.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | ENST00000373019.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | ENST00000373019.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | ENST00000373019.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | ENST00000373019.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | ENST00000373019.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | ENST00000460925.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | ENST00000460925.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | ENST00000460925.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | ENST00000460925.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | ENST00000460925.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | ENST00000460925.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | ENST00000460925.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | ENST00000461869.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | ENST00000461869.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | ENST00000461869.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | ENST00000461869.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | ENST00000461869.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | ENST00000461869.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | ENST00000461869.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | ENST00000461869.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | ENST00000461869.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | ENST00000462258.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | ENST00000462258.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | ENST00000462258.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | ENST00000462258.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | ENST00000462258.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | ENST00000462258.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | ENST00000462258.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | ENST00000470585.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | ENST00000470585.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | ENST00000470585.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | ENST00000470585.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | ENST00000470585.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | ENST00000470585.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | ENST00000470585.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | ENST00000470585.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | ENST00000487062.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | ENST00000487062.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | ENST00000487062.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | ENST00000487062.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | ENST00000488934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | ENST00000488934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | ENST00000488934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | ENST00000489537.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | ENST00000489537.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | ENST00000489537.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | ENST00000489537.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | ENST00000489537.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | ENST00000489537.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | ENST00000489537.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | ENST00000489537.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | ENST00000489537.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | ENST00000489537.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | ENST00000489537.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | NM_006802.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | NM_006802.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | NM_006802.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | NM_006802.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | NM_006802.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | NM_006802.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | NM_006802.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | NM_006802.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | NM_006802.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | NM_006802.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | NM_006802.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | NM_006802.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | NM_006802.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | NM_006802.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | NM_006802.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | NM_006802.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | NM_006802.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | NM_006802.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | NM_006802.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | NM_006802.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | NM_006802.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013760.hg.1 | NM_006802.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100202775.hg.1 | ENST00000373019.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100202775.hg.1 | ENST00000460925.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100202775.hg.1 | ENST00000489537.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100202775.hg.1 | NM_006802.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100202775.hg.1 | ENST00000373019.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100202775.hg.1 | ENST00000460925.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100202775.hg.1 | ENST00000489537.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100202775.hg.1 | NM_006802.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100202775.hg.1 | TC0100013760.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | ENST00000373019.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | ENST00000373019.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | ENST00000373019.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | ENST00000373019.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | ENST00000373019.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | ENST00000373019.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | ENST00000373019.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | ENST00000373019.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | ENST00000373019.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | ENST00000373019.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | ENST00000373019.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | ENST00000373019.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | ENST00000373019.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | ENST00000373019.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | ENST00000373019.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | ENST00000373019.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | ENST00000373019.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | ENST00000373019.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | ENST00000373019.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | ENST00000373019.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | ENST00000373019.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | ENST00000373019.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | ENST00000373019.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | ENST00000460925.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | ENST00000460925.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | ENST00000460925.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | ENST00000460925.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | ENST00000460925.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | ENST00000460925.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | ENST00000460925.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | ENST00000461869.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | ENST00000461869.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | ENST00000461869.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | ENST00000461869.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | ENST00000461869.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | ENST00000461869.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | ENST00000461869.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | ENST00000461869.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | ENST00000461869.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | ENST00000462258.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | ENST00000462258.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | ENST00000462258.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | ENST00000462258.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | ENST00000462258.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | ENST00000462258.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | ENST00000462258.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | ENST00000470585.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | ENST00000470585.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | ENST00000470585.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | ENST00000470585.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | ENST00000470585.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | ENST00000470585.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | ENST00000470585.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | ENST00000470585.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | ENST00000487062.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | ENST00000487062.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | ENST00000487062.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | ENST00000487062.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | ENST00000488934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | ENST00000488934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | ENST00000488934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | ENST00000489537.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | ENST00000489537.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | ENST00000489537.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | ENST00000489537.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | ENST00000489537.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | ENST00000489537.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | ENST00000489537.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | ENST00000489537.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | ENST00000489537.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | ENST00000489537.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | ENST00000489537.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | NM_006802.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | NM_006802.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | NM_006802.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | NM_006802.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | NM_006802.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | NM_006802.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | NM_006802.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | NM_006802.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | NM_006802.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | NM_006802.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | NM_006802.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | NM_006802.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | NM_006802.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | NM_006802.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | NM_006802.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | NM_006802.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | NM_006802.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | NM_006802.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | NM_006802.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | NM_006802.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | NM_006802.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013760.hg.1 | NM_006802.3 | Refseq |

SF3A3
Genomic Map