Assay Details
| Gene Symbol: CPT1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_108072.1 | 2 - 3 | 347 | 55 | ||
| XM_005258505.2 | XP_005258562.1 | 2 - 3 | 343 | 55 | ||
| XM_006723009.3 | XP_006723072.1 | 1 - 2 | 148 | 55 | ||
| XM_017026268.1 | XP_016881757.1 | 1 - 2 | 148 | 55 | ||
| XM_017026271.1 | XP_016881760.1 | 1 - 2 | 148 | 55 | ||
| XM_017026273.1 | XP_016881762.1 | 2 - 3 | 327 | 55 | ||
| GenBank mRNA | AK096516.1 | - | 2 - 3 | 343 | 55 | |
| AK299866.1 | - | 1 - 2 | 137 | 55 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK096516.1, AK299866.1, XM_005258505.2, XM_006723009.3, XM_017026268.1, XM_017026271.1, XM_017026273.1 transcripts.
Set Membership: |
|

CPT1C
Genomic Map