Assay Details
| Gene Symbol: SLC39A7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077516.1 | NP_001070984.1 | 3 - 4 | 695 | 59 | |
| NM_001288777.1 | NP_001275706.1 | 3 - 4 | 411 | 59 | ||
| NM_006979.2 | NP_008910.2 | 2 - 3 | 998 | 59 | ||
| GenBank mRNA | AK225282.1 | - | 2 - 3 | 922 | 59 | |
| AK301126.1 | - | 4 - 5 | 597 | 59 | ||
| BC000645.2 | - | 3 - 4 | 633 | 3345970 | 59 | |
| D82060.1 | - | 2 - 3 | 875 | 59 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06000412.hg.1, TC6_cox_hap2000102.hg.1, TC6_dbb_hap3000094.hg.1, TC6_mann_hap4000086.hg.1, TC6_mcf_hap5000087.hg.1, TC6_qbl_hap6000093.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR06004837.hg.1 | TC06000412.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06004837.hg.1 | ENST00000374675.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06004837.hg.1 | ENST00000374677.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06004837.hg.1 | ENST00000445037.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06004837.hg.1 | ENST00000446283.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06004837.hg.1 | ENST00000496894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06004837.hg.1 | NM_001077516.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06004837.hg.1 | NM_006979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3001885.hg.1 | TC6_dbb_hap3000094.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3001885.hg.1 | NM_001077516.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3001885.hg.1 | NM_006979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06004836.hg.1 | TC06000412.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06004836.hg.1 | ENST00000374675.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06004836.hg.1 | ENST00000374677.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06004836.hg.1 | ENST00000445037.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06004836.hg.1 | ENST00000446283.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06004836.hg.1 | ENST00000496894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06004836.hg.1 | NM_001077516.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06004836.hg.1 | NM_006979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007684.hg.1 | ENST00000374675.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007684.hg.1 | ENST00000374675.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007684.hg.1 | ENST00000374675.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007684.hg.1 | ENST00000374675.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007684.hg.1 | ENST00000374675.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007684.hg.1 | ENST00000374675.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007684.hg.1 | ENST00000374675.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007684.hg.1 | ENST00000374675.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007684.hg.1 | ENST00000374675.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007684.hg.1 | ENST00000374675.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007684.hg.1 | ENST00000374675.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007684.hg.1 | ENST00000374677.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007684.hg.1 | ENST00000374677.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007684.hg.1 | ENST00000374677.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007684.hg.1 | ENST00000374677.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007684.hg.1 | ENST00000374677.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007684.hg.1 | ENST00000374677.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007684.hg.1 | ENST00000374677.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007684.hg.1 | ENST00000374677.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007684.hg.1 | ENST00000374677.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007684.hg.1 | ENST00000374677.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007684.hg.1 | ENST00000374677.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007684.hg.1 | ENST00000444757.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007684.hg.1 | ENST00000444757.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007684.hg.1 | ENST00000444757.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007684.hg.1 | ENST00000444757.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007684.hg.1 | ENST00000444757.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007684.hg.1 | ENST00000444757.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007684.hg.1 | ENST00000444757.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007684.hg.1 | ENST00000463972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007684.hg.1 | ENST00000463972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007684.hg.1 | ENST00000463972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007684.hg.1 | ENST00000463972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007684.hg.1 | ENST00000463972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007684.hg.1 | ENST00000617885.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007684.hg.1 | ENST00000617885.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007684.hg.1 | ENST00000617885.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007684.hg.1 | ENST00000617885.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007684.hg.1 | ENST00000617885.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007684.hg.1 | ENST00000617885.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007684.hg.1 | ENST00000617885.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007684.hg.1 | ENST00000617885.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007684.hg.1 | ENST00000617885.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007684.hg.1 | ENST00000617885.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007684.hg.1 | NM_001288777.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007684.hg.1 | NM_001288777.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007684.hg.1 | NM_001288777.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007684.hg.1 | NM_001288777.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007684.hg.1 | NM_001288777.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007684.hg.1 | NM_001288777.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007684.hg.1 | NM_001288777.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007684.hg.1 | NM_001288777.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007684.hg.1 | NM_001288777.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007684.hg.1 | NM_001288777.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007684.hg.1 | NM_006979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007684.hg.1 | NM_006979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007684.hg.1 | NM_006979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007684.hg.1 | NM_006979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007684.hg.1 | NM_006979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007684.hg.1 | NM_006979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007684.hg.1 | NM_006979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007684.hg.1 | NM_006979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007684.hg.1 | NM_006979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007684.hg.1 | NM_006979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007684.hg.1 | NM_006979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007684.hg.1 | NM_006979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2001902.hg.1 | TC6_cox_hap2000102.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2001902.hg.1 | NM_001077516.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2001902.hg.1 | NM_006979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4001513.hg.1 | TC6_mann_hap4000086.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4001513.hg.1 | NM_001077516.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4001513.hg.1 | NM_006979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5001680.hg.1 | TC6_mcf_hap5000087.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5001680.hg.1 | NM_001077516.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5001680.hg.1 | NM_006979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6001866.hg.1 | TC6_qbl_hap6000093.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6001866.hg.1 | NM_001077516.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6001866.hg.1 | NM_006979.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007684.hg.1 | ENST00000374675.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007684.hg.1 | ENST00000374675.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007684.hg.1 | ENST00000374675.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007684.hg.1 | ENST00000374675.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007684.hg.1 | ENST00000374675.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007684.hg.1 | ENST00000374675.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007684.hg.1 | ENST00000374675.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007684.hg.1 | ENST00000374675.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007684.hg.1 | ENST00000374675.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007684.hg.1 | ENST00000374675.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007684.hg.1 | ENST00000374675.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007684.hg.1 | ENST00000374677.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007684.hg.1 | ENST00000374677.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007684.hg.1 | ENST00000374677.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007684.hg.1 | ENST00000374677.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007684.hg.1 | ENST00000374677.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007684.hg.1 | ENST00000374677.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007684.hg.1 | ENST00000374677.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007684.hg.1 | ENST00000374677.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007684.hg.1 | ENST00000374677.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007684.hg.1 | ENST00000374677.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007684.hg.1 | ENST00000374677.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007684.hg.1 | ENST00000444757.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007684.hg.1 | ENST00000444757.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007684.hg.1 | ENST00000444757.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007684.hg.1 | ENST00000444757.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007684.hg.1 | ENST00000444757.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007684.hg.1 | ENST00000444757.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007684.hg.1 | ENST00000444757.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007684.hg.1 | ENST00000463972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007684.hg.1 | ENST00000463972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007684.hg.1 | ENST00000463972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007684.hg.1 | ENST00000463972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007684.hg.1 | ENST00000463972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007684.hg.1 | ENST00000617885.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007684.hg.1 | ENST00000617885.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007684.hg.1 | ENST00000617885.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007684.hg.1 | ENST00000617885.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007684.hg.1 | ENST00000617885.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007684.hg.1 | ENST00000617885.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007684.hg.1 | ENST00000617885.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007684.hg.1 | ENST00000617885.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007684.hg.1 | ENST00000617885.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007684.hg.1 | ENST00000617885.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007684.hg.1 | NM_001288777.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007684.hg.1 | NM_001288777.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007684.hg.1 | NM_001288777.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007684.hg.1 | NM_001288777.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007684.hg.1 | NM_001288777.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007684.hg.1 | NM_001288777.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007684.hg.1 | NM_001288777.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007684.hg.1 | NM_001288777.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007684.hg.1 | NM_001288777.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007684.hg.1 | NM_001288777.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007684.hg.1 | NM_006979.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007684.hg.1 | NM_006979.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007684.hg.1 | NM_006979.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007684.hg.1 | NM_006979.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007684.hg.1 | NM_006979.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007684.hg.1 | NM_006979.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007684.hg.1 | NM_006979.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007684.hg.1 | NM_006979.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007684.hg.1 | NM_006979.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007684.hg.1 | NM_006979.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007684.hg.1 | NM_006979.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007684.hg.1 | NM_006979.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600153105.hg.1 | ENST00000374675.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600153105.hg.1 | ENST00000374677.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600153105.hg.1 | ENST00000444757.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600153105.hg.1 | ENST00000617885.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600153105.hg.1 | NM_001288777.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600153105.hg.1 | NM_006979.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600153105.hg.1 | ENST00000374675.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600153105.hg.1 | ENST00000374677.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600153105.hg.1 | ENST00000444757.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600153105.hg.1 | ENST00000617885.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600153105.hg.1 | NM_001288777.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600153105.hg.1 | NM_006979.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600153105.hg.1 | TC0600007684.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 179975 | - | - |

SLC39A7
Genomic Map