Assay Details
| Gene Symbol: AMPD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000036.2 | NP_000027.2 | 12 - 13 | 1830 | 68 | |
| NM_001172626.1 | NP_001166097.1 | 11 - 12 | 1818 | 68 | ||
| GenBank mRNA | AK097077.1 | - | 11 - 12 | 1817 | 68 | |
| AK291349.1 | - | 12 - 13 | 1734 | 68 | ||
| M60092.1 | - | 12 - 13 | 1767 | 68 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01003013.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01046989.hg.1 | TC01003013.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046989.hg.1 | ENST00000353928.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046989.hg.1 | ENST00000369538.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046989.hg.1 | ENST00000520113.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046989.hg.1 | NM_000036.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046989.hg.1 | NM_001172626.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | ENST00000369538.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | ENST00000369538.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | ENST00000369538.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | ENST00000369538.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | ENST00000369538.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | ENST00000369538.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | ENST00000369538.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | ENST00000369538.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | ENST00000369538.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | ENST00000369538.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | ENST00000369538.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | ENST00000369538.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | ENST00000369538.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | ENST00000369538.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | ENST00000369538.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | ENST00000369538.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | ENST00000485564.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | ENST00000485564.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | ENST00000485564.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | ENST00000520113.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | ENST00000520113.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | ENST00000520113.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | ENST00000520113.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | ENST00000520113.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | ENST00000520113.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | ENST00000520113.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | ENST00000520113.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | ENST00000520113.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | ENST00000520113.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | ENST00000520113.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | ENST00000520113.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | ENST00000520113.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | ENST00000520113.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | ENST00000520113.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | NM_000036.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | NM_000036.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | NM_000036.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | NM_000036.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | NM_000036.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | NM_000036.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | NM_000036.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | NM_000036.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | NM_000036.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | NM_000036.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | NM_000036.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | NM_000036.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | NM_000036.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | NM_000036.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | NM_000036.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | NM_000036.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | NM_001172626.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | NM_001172626.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | NM_001172626.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | NM_001172626.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | NM_001172626.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | NM_001172626.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | NM_001172626.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | NM_001172626.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | NM_001172626.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | NM_001172626.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | NM_001172626.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | NM_001172626.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | NM_001172626.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | NM_001172626.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | NM_001172626.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | NM_001172626.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | ENST00000369538.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | ENST00000369538.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | ENST00000369538.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | ENST00000369538.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | ENST00000369538.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | ENST00000369538.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | ENST00000369538.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | ENST00000369538.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | ENST00000369538.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | ENST00000369538.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | ENST00000369538.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | ENST00000369538.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | ENST00000369538.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | ENST00000369538.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | ENST00000369538.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | ENST00000369538.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | ENST00000485564.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | ENST00000485564.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | ENST00000485564.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | ENST00000520113.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | ENST00000520113.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | ENST00000520113.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | ENST00000520113.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | ENST00000520113.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | ENST00000520113.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | ENST00000520113.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | ENST00000520113.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | ENST00000520113.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | ENST00000520113.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | ENST00000520113.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | ENST00000520113.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | ENST00000520113.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | ENST00000520113.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | ENST00000520113.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | NM_000036.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | NM_000036.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | NM_000036.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | NM_000036.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | NM_000036.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | NM_000036.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | NM_000036.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | NM_000036.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | NM_000036.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | NM_000036.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | NM_000036.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | NM_000036.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | NM_000036.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | NM_000036.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | NM_000036.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | NM_000036.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | NM_001172626.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | NM_001172626.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | NM_001172626.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | NM_001172626.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | NM_001172626.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | NM_001172626.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | NM_001172626.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | NM_001172626.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | NM_001172626.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | NM_001172626.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | NM_001172626.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | NM_001172626.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | NM_001172626.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | NM_001172626.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | NM_001172626.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | NM_001172626.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100214687.hg.1 | ENST00000369538.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100214687.hg.1 | ENST00000520113.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100214687.hg.1 | NM_000036.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100214687.hg.1 | NM_001172626.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100214687.hg.1 | ENST00000369538.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100214687.hg.1 | ENST00000520113.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100214687.hg.1 | NM_000036.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100214687.hg.1 | NM_001172626.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100214687.hg.1 | TC0100015350.hg.1 | Clariom D TC |

AMPD1
Genomic Map