Assay Details
| Gene Symbol: FANCG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004629.1 | NP_004620.1 | 11 - 12 | 1970 | 56 | |
| GenBank mRNA | AJ007669.1 | - | 11 - 12 | 1970 | 56 | |
| AK311348.1 | - | 10 - 11 | 1444 | 56 | ||
| AK312987.1 | - | 11 - 12 | 1895 | 56 | ||
| BC000032.2 | - | 11 - 12 | 1616 | 3507258 | 56 | |
| BC011623.2 | - | 11 - 12 | 1743 | 4053144 | 56 | |
| BT009813.1 | - | 11 - 12 | 1478 | 56 | ||
| KU159422.1 | - | 11 - 12 | 1983 | 56 | ||
| KU178053.1 | - | 11 - 12 | 1478 | 56 | ||
| KU178054.1 | - | 11 - 12 | 1460 | 56 | ||
| KU178055.1 | - | 11 - 12 | 1478 | 56 | ||
| KU178056.1 | - | 8 - 9 | 1052 | 56 | ||
| U70310.1 | - | 11 - 12 | 1612 | 56 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09001049.hg.1
Set Membership: |
|

FANCG
Genomic Map