| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000378643.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000378643.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000378643.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000378643.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000378643.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000378643.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000378643.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000378643.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000378643.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000378643.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000378643.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000378643.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000378643.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000378643.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000378643.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000378643.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000378643.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000378643.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000378643.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000378643.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000378643.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000378643.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000378643.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000378643.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000425676.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000425676.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000425676.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000425676.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000425676.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000425676.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000425676.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000425676.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000425676.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000425676.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000425676.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000425676.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000425676.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000425676.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000425676.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000425676.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000425676.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000425676.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000425676.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000448890.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000448890.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000448890.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000448890.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000448890.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000448890.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000448890.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000448890.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000461149.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000461149.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000461149.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000462124.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000462124.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000462124.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000462124.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000462124.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000462124.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000474894.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000474894.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000474894.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000474894.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000476212.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000476212.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000476212.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000476212.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000481254.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000481254.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000481254.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000481254.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
ENST00000481254.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
NM_004629.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
NM_004629.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
NM_004629.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
NM_004629.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
NM_004629.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
NM_004629.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
NM_004629.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
NM_004629.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
NM_004629.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
NM_004629.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
NM_004629.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
NM_004629.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
NM_004629.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
NM_004629.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
NM_004629.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
NM_004629.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
NM_004629.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
NM_004629.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
NM_004629.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
NM_004629.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
NM_004629.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
NM_004629.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
NM_004629.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009930.hg.1 |
NM_004629.1 |
Refseq |