Assay Details
| Gene Symbol: KLC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177417.2 | NP_803136.2 | 11 - 12 | 1493 | 88 | |
| GenBank mRNA | AK092481.1 | - | 10 - 11 | 1520 | 88 | |
| BC062998.1 | - | 11 - 12 | 1480 | 5172274 | 88 | |
| BC073841.1 | - | 11 - 12 | 1469 | 5190809 | 88 | |
| BC126418.1 | - | 10 - 11 | 1509 | 8992134 | 88 | |
| BC133037.1 | - | 10 - 11 | 1509 | 40147059 | 88 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19000645.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19009034.hg.1 | ENST00000391946.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19009034.hg.1 | ENST00000470402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19009034.hg.1 | NM_177417.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19009034.hg.1 | TC19000645.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000391946.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000391946.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000391946.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000391946.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000391946.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000391946.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000391946.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000391946.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000391946.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000391946.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000391946.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000391946.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000391946.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000391946.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000391946.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000391946.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000391946.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000391946.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000470402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000470402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000470402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000470402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000470402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000470402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000470402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000470402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000470402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000470402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000470402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000470402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000470402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000470402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000470402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000470402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000470402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000470402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000494686.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000494686.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000494686.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000494686.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000494686.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000585434.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000585434.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000585434.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000585434.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000585434.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000585434.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000585434.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000585434.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000585434.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000585434.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000585434.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000585434.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000585434.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000585434.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000585434.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000585434.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000585434.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000585434.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000589373.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000589373.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000589373.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000589373.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000589373.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000589373.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000589373.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000589373.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000589373.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000589373.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000589373.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000589373.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000589837.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000589837.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000589837.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000589837.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000589837.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000589837.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000589837.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000590063.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000590063.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000590063.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | ENST00000590063.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | NM_177417.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | NM_177417.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | NM_177417.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | NM_177417.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | NM_177417.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | NM_177417.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | NM_177417.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | NM_177417.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | NM_177417.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | NM_177417.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | NM_177417.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | NM_177417.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | NM_177417.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | NM_177417.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | NM_177417.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | NM_177417.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | NM_177417.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008321.hg.1 | NM_177417.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000391946.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000391946.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000391946.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000391946.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000391946.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000391946.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000391946.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000391946.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000391946.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000391946.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000391946.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000391946.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000391946.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000391946.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000391946.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000391946.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000391946.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000391946.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000470402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000470402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000470402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000470402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000470402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000470402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000470402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000470402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000470402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000470402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000470402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000470402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000470402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000470402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000470402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000470402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000470402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000470402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000494686.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000494686.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000494686.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000494686.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000494686.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000585434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000585434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000585434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000585434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000585434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000585434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000585434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000585434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000585434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000585434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000585434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000585434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000585434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000585434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000585434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000585434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000585434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000585434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000589373.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000589373.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000589373.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000589373.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000589373.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000589373.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000589373.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000589373.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000589373.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000589373.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000589373.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000589373.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000589837.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000589837.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000589837.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000589837.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000589837.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000589837.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000589837.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000590063.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000590063.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000590063.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | ENST00000590063.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | NM_177417.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | NM_177417.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | NM_177417.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | NM_177417.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | NM_177417.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | NM_177417.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | NM_177417.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | NM_177417.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | NM_177417.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | NM_177417.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | NM_177417.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | NM_177417.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | NM_177417.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | NM_177417.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | NM_177417.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | NM_177417.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | NM_177417.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008321.hg.1 | NM_177417.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900163844.hg.1 | ENST00000391946.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900163844.hg.1 | ENST00000470402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900163844.hg.1 | ENST00000585434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900163844.hg.1 | NM_177417.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900163844.hg.1 | TC1900008321.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900163844.hg.1 | ENST00000391946.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900163844.hg.1 | ENST00000470402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900163844.hg.1 | ENST00000585434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900163844.hg.1 | NM_177417.2 | Refseq |

KLC3
Genomic Map