Assay Details
| Gene Symbol: LRCH3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032773.3 | NP_116162.1 | 15 - 16 | 1767 | 89 | |
| XM_005269362.2 | XP_005269419.1 | 15 - 16 | 2138 | 89 | ||
| XM_006713791.3 | XP_006713854.1 | 15 - 16 | 2138 | 89 | ||
| XM_017007350.1 | XP_016862839.1 | 15 - 16 | 2138 | 89 | ||
| GenBank mRNA | AK055819.1 | - | 10 - 11 | 873 | 89 | |
| AK293745.1 | - | 15 - 16 | 1756 | 89 | ||
| AK300466.1 | - | 12 - 13 | 1267 | 89 | ||
| BC007504.2 | - | 15 - 16 | 1719 | 2960711 | 89 | |
More Information
Set Membership: |
|

LRCH3
Genomic Map