Assay Details
| Gene Symbol: CAMK1G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020439.2 | NP_065172.1 | 10 - 11 | 1014 | 96 | |
| XM_017001866.1 | XP_016857355.1 | 10 - 11 | 1157 | 96 | ||
| XM_017001867.1 | XP_016857356.1 | 10 - 11 | 1008 | 96 | ||
| GenBank mRNA | AF428261.1 | - | 10 - 11 | 984 | 96 | |
| AK095713.1 | - | 10 - 11 | 1157 | 96 | ||
| AK289675.1 | - | 10 - 11 | 997 | 96 | ||
| AL049688.1 | - | 9 - 10 | 929 | 96 | ||
| AY212935.1 | - | 9 - 10 | 920 | 96 | ||
| BC032787.1 | - | 10 - 11 | 981 | 5179957 | 96 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01001748.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000423146.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000423146.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000423146.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000423146.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000423146.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000423146.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000423146.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000423146.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000423146.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000494990.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000494990.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000494990.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000494990.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000494990.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000494990.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01027374.hg.1 | TC01001748.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01027374.hg.1 | ENST00000009105.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01027374.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01027374.hg.1 | ENST00000494990.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01027374.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01027375.hg.1 | TC01001748.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01027375.hg.1 | ENST00000009105.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01027375.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01027375.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100184332.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100184332.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100184332.hg.1 | ENST00000494990.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100184332.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100184332.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100184332.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100184332.hg.1 | ENST00000494990.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100184332.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100184332.hg.1 | TC0100011451.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100184333.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100184333.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100184333.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100184333.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100184333.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100184333.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100184333.hg.1 | TC0100011451.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000423146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000423146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000423146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000423146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000423146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000423146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000423146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000423146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000423146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000494990.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000494990.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000494990.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000494990.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000494990.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000494990.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | NM_020439.2 | Refseq |

CAMK1G
Genomic Map