Assay Details
| Gene Symbol: PPM1F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014634.3 | NP_055449.1 | 6 - 7 | 1034 | 64 | |
| XM_011530556.1 | XP_011528858.1 | 5 - 6 | 765 | 64 | ||
| GenBank mRNA | AF305840.1 | - | 5 - 6 | 997 | 64 | |
| AF520615.1 | - | 5 - 6 | 887 | 64 | ||
| AK126377.1 | - | 4 - 5 | 1444 | 64 | ||
| AK291628.1 | - | 6 - 7 | 1004 | 64 | ||
| AK294557.1 | - | 4 - 5 | 891 | 64 | ||
| BC071989.1 | - | 6 - 7 | 976 | 5421075 | 64 | |
| D13640.1 | - | 6 - 7 | 993 | 64 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22000548.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of protein kinase activity
positive regulation of gene expression
positive regulation of epithelial cell migration
positive regulation of cell-substrate adhesion
histone dephosphorylation
negative regulation of peptidyl-serine phosphorylation
cellular response to drug
peptidyl-threonine dephosphorylation
positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
negative regulation of protein kinase activity by regulation of protein phosphorylation
negative regulation of transcription, DNA-templated
positive regulation of growth
positive regulation of chemotaxis
positive regulation of stress fiber assembly
positive regulation of focal adhesion assembly
intrinsic apoptotic signaling pathway
protein serine/threonine phosphatase activity
protein binding
phosphatase activity
calmodulin-dependent protein phosphatase activity
metal ion binding
negative regulation of protein kinase activity
positive regulation of gene expression
positive regulation of epithelial cell migration
positive regulation of cell-substrate adhesion
histone dephosphorylation
negative regulation of peptidyl-serine phosphorylation
cellular response to drug
peptidyl-threonine dephosphorylation
positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
negative regulation of protein kinase activity by regulation of protein phosphorylation
negative regulation of transcription, DNA-templated
positive regulation of growth
positive regulation of chemotaxis
positive regulation of stress fiber assembly
positive regulation of focal adhesion assembly
intrinsic apoptotic signaling pathway
protein serine/threonine phosphatase activity
protein binding
phosphatase activity
calmodulin-dependent protein phosphatase activity
metal ion binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2200008110.hg.1 | ENST00000263212.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008110.hg.1 | ENST00000263212.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008110.hg.1 | ENST00000263212.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008110.hg.1 | ENST00000263212.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008110.hg.1 | ENST00000263212.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008110.hg.1 | ENST00000263212.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008110.hg.1 | ENST00000263212.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008110.hg.1 | ENST00000263212.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008110.hg.1 | ENST00000263212.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008110.hg.1 | ENST00000263212.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008110.hg.1 | ENST00000263212.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008110.hg.1 | ENST00000263212.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008110.hg.1 | ENST00000263212.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008110.hg.1 | ENST00000397495.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008110.hg.1 | ENST00000397495.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008110.hg.1 | ENST00000397495.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008110.hg.1 | ENST00000397495.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008110.hg.1 | ENST00000397495.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008110.hg.1 | ENST00000397495.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008110.hg.1 | ENST00000397495.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008110.hg.1 | ENST00000397495.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008110.hg.1 | ENST00000397495.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008110.hg.1 | ENST00000397495.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008110.hg.1 | ENST00000397495.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008110.hg.1 | ENST00000407142.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008110.hg.1 | ENST00000407142.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008110.hg.1 | ENST00000407142.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008110.hg.1 | ENST00000407142.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008110.hg.1 | ENST00000407142.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008110.hg.1 | ENST00000407142.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008110.hg.1 | ENST00000407142.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008110.hg.1 | ENST00000407142.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008110.hg.1 | ENST00000407142.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008110.hg.1 | ENST00000407142.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008110.hg.1 | ENST00000407142.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008110.hg.1 | ENST00000407142.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008110.hg.1 | ENST00000424647.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008110.hg.1 | ENST00000424647.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008110.hg.1 | ENST00000424647.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008110.hg.1 | ENST00000445205.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008110.hg.1 | ENST00000445205.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008110.hg.1 | ENST00000445205.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008110.hg.1 | ENST00000445205.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008110.hg.1 | ENST00000445205.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008110.hg.1 | ENST00000484588.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008110.hg.1 | ENST00000484588.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008110.hg.1 | ENST00000484588.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008110.hg.1 | ENST00000486259.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008110.hg.1 | ENST00000486259.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008110.hg.1 | ENST00000496143.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008110.hg.1 | ENST00000496143.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008110.hg.1 | ENST00000496143.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008110.hg.1 | ENST00000497072.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008110.hg.1 | ENST00000497072.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008110.hg.1 | ENST00000623652.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008110.hg.1 | ENST00000623652.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008110.hg.1 | ENST00000623652.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008110.hg.1 | ENST00000623652.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008110.hg.1 | NM_014634.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008110.hg.1 | NM_014634.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008110.hg.1 | NM_014634.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008110.hg.1 | NM_014634.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008110.hg.1 | NM_014634.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008110.hg.1 | NM_014634.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008110.hg.1 | NM_014634.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008110.hg.1 | NM_014634.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008110.hg.1 | NM_014634.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008110.hg.1 | NM_014634.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008110.hg.1 | NM_014634.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008110.hg.1 | NM_014634.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008110.hg.1 | NM_014634.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008110.hg.1 | NM_014634.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22009908.hg.1 | ENST00000263212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22009908.hg.1 | ENST00000397495.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22009908.hg.1 | ENST00000406981.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22009908.hg.1 | ENST00000407142.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22009908.hg.1 | ENST00000538191.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22009908.hg.1 | NM_014634.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22009908.hg.1 | TC22000548.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC2200008110.hg.1 | ENST00000263212.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008110.hg.1 | ENST00000263212.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008110.hg.1 | ENST00000263212.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008110.hg.1 | ENST00000263212.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008110.hg.1 | ENST00000263212.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008110.hg.1 | ENST00000263212.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008110.hg.1 | ENST00000263212.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008110.hg.1 | ENST00000263212.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008110.hg.1 | ENST00000263212.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008110.hg.1 | ENST00000263212.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008110.hg.1 | ENST00000263212.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008110.hg.1 | ENST00000263212.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008110.hg.1 | ENST00000263212.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008110.hg.1 | ENST00000397495.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008110.hg.1 | ENST00000397495.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008110.hg.1 | ENST00000397495.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008110.hg.1 | ENST00000397495.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008110.hg.1 | ENST00000397495.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008110.hg.1 | ENST00000397495.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008110.hg.1 | ENST00000397495.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008110.hg.1 | ENST00000397495.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008110.hg.1 | ENST00000397495.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008110.hg.1 | ENST00000397495.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008110.hg.1 | ENST00000397495.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008110.hg.1 | ENST00000407142.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008110.hg.1 | ENST00000407142.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008110.hg.1 | ENST00000407142.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008110.hg.1 | ENST00000407142.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008110.hg.1 | ENST00000407142.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008110.hg.1 | ENST00000407142.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008110.hg.1 | ENST00000407142.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008110.hg.1 | ENST00000407142.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008110.hg.1 | ENST00000407142.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008110.hg.1 | ENST00000407142.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008110.hg.1 | ENST00000407142.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008110.hg.1 | ENST00000407142.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008110.hg.1 | ENST00000424647.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008110.hg.1 | ENST00000424647.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008110.hg.1 | ENST00000424647.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008110.hg.1 | ENST00000445205.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008110.hg.1 | ENST00000445205.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008110.hg.1 | ENST00000445205.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008110.hg.1 | ENST00000445205.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008110.hg.1 | ENST00000445205.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008110.hg.1 | ENST00000484588.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008110.hg.1 | ENST00000484588.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008110.hg.1 | ENST00000484588.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008110.hg.1 | ENST00000486259.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008110.hg.1 | ENST00000486259.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008110.hg.1 | ENST00000496143.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008110.hg.1 | ENST00000496143.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008110.hg.1 | ENST00000496143.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008110.hg.1 | ENST00000497072.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008110.hg.1 | ENST00000497072.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008110.hg.1 | ENST00000623652.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008110.hg.1 | ENST00000623652.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008110.hg.1 | ENST00000623652.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008110.hg.1 | ENST00000623652.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008110.hg.1 | NM_014634.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008110.hg.1 | NM_014634.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008110.hg.1 | NM_014634.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008110.hg.1 | NM_014634.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008110.hg.1 | NM_014634.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008110.hg.1 | NM_014634.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008110.hg.1 | NM_014634.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008110.hg.1 | NM_014634.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008110.hg.1 | NM_014634.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008110.hg.1 | NM_014634.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008110.hg.1 | NM_014634.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008110.hg.1 | NM_014634.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008110.hg.1 | NM_014634.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008110.hg.1 | NM_014634.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200158024.hg.1 | TC2200008110.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200158024.hg.1 | ENST00000263212.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158024.hg.1 | ENST00000397495.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158024.hg.1 | ENST00000407142.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158024.hg.1 | ENST00000484588.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158024.hg.1 | ENST00000496143.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158024.hg.1 | NM_014634.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200158024.hg.1 | ENST00000263212.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158024.hg.1 | ENST00000397495.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158024.hg.1 | ENST00000407142.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158024.hg.1 | ENST00000484588.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158024.hg.1 | ENST00000496143.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158024.hg.1 | NM_014634.3 | Refseq |

PPM1F
Genomic Map