Assay Details
| Gene Symbol: SNRK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100594.1 | NP_001094064.1 | 3 - 4 | 987 | 124 | |
| NM_017719.4 | NP_060189.3 | 4 - 5 | 1049 | 124 | ||
| XM_005265245.3 | XP_005265302.1 | 5 - 6 | 1099 | 124 | ||
| XM_011533861.2 | XP_011532163.1 | 2 - 3 | 240 | 124 | ||
| GenBank mRNA | AK291360.1 | - | 5 - 6 | 1100 | 124 | |
| AK295187.1 | - | 2 - 3 | 218 | 124 | ||
| AK314376.1 | - | 3 - 4 | 872 | 124 | ||
| BC071567.1 | - | 3 - 4 | 961 | 4375623 | 124 | |
| D43636.2 | - | 2 - 3 | 721 | 124 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03000229.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03004645.hg.1 | TC03000229.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03004645.hg.1 | ENST00000296088.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03004645.hg.1 | ENST00000429705.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03004645.hg.1 | ENST00000437827.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03004645.hg.1 | ENST00000454177.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03004645.hg.1 | NM_001100594.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03004645.hg.1 | NM_017719.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000296088.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000296088.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000296088.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000296088.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000296088.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000296088.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000296088.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000296088.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000296088.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000296088.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000296088.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000296088.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000296088.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000429705.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000429705.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000429705.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000429705.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000429705.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000429705.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000429705.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000429705.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000429705.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000429705.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000429705.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000429705.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000437827.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000437827.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000437827.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000437827.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000437827.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000437827.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000437827.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000454177.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000454177.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000454177.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000454177.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000454177.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000454177.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000454177.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000454177.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000454177.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000454177.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000454177.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000454177.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000454177.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000454177.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000462810.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000462810.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000462810.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000462810.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000468628.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000468628.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000468628.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000481892.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000481892.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000481892.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000484791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000484791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000484791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | NM_001100594.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | NM_001100594.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | NM_001100594.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | NM_001100594.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | NM_001100594.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | NM_001100594.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | NM_001100594.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | NM_001100594.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | NM_001100594.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | NM_001100594.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | NM_001100594.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | NM_001100594.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | NM_001100594.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | NM_017719.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | NM_017719.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | NM_017719.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | NM_017719.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | NM_017719.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | NM_017719.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | NM_017719.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | NM_017719.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | NM_017719.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | NM_017719.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | NM_017719.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | NM_017719.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | NM_017719.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | NM_017719.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000296088.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000296088.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000296088.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000296088.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000296088.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000296088.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000296088.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000296088.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000296088.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000296088.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000296088.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000296088.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000296088.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000429705.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000429705.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000429705.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000429705.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000429705.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000429705.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000429705.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000429705.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000429705.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000429705.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000429705.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000429705.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000437827.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000437827.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000437827.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000437827.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000437827.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000437827.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000437827.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000454177.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000454177.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000454177.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000454177.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000454177.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000454177.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000454177.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000454177.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000454177.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000454177.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000454177.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000454177.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000454177.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000454177.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000462810.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000462810.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000462810.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000462810.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000468628.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000468628.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000468628.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000481892.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000481892.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000481892.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000484791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000484791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000484791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | NM_001100594.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | NM_001100594.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | NM_001100594.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | NM_001100594.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | NM_001100594.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | NM_001100594.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | NM_001100594.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | NM_001100594.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | NM_001100594.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | NM_001100594.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | NM_001100594.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | NM_001100594.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | NM_001100594.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | NM_017719.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | NM_017719.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | NM_017719.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | NM_017719.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | NM_017719.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | NM_017719.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | NM_017719.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | NM_017719.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | NM_017719.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | NM_017719.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | NM_017719.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | NM_017719.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | NM_017719.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | NM_017719.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300150671.hg.1 | ENST00000296088.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300150671.hg.1 | ENST00000429705.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300150671.hg.1 | ENST00000437827.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300150671.hg.1 | ENST00000454177.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300150671.hg.1 | ENST00000484791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300150671.hg.1 | NM_001100594.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300150671.hg.1 | NM_017719.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300150671.hg.1 | ENST00000296088.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300150671.hg.1 | ENST00000429705.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300150671.hg.1 | ENST00000437827.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300150671.hg.1 | ENST00000454177.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300150671.hg.1 | ENST00000484791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300150671.hg.1 | NM_001100594.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300150671.hg.1 | NM_017719.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300150671.hg.1 | TC0300007189.hg.1 | Clariom D TC |

SNRK
Genomic Map