Assay Details
| Gene Symbol: BAIAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006340.2 | NP_006331.1 | 14 - 15 | 1681 | 80 | |
| XM_005256945.1 | XP_005257002.1 | 15 - 16 | 1780 | 80 | ||
| XM_006721636.1 | XP_006721699.1 | 15 - 16 | 1783 | 80 | ||
| GenBank mRNA | AB015020.1 | - | 14 - 15 | 1666 | 80 | |
| AK056888.1 | - | 10 - 11 | 1853 | 80 | ||
More Information
Set Membership: |
|

BAIAP2
Genomic Map