Assay Details
| Gene Symbol: SPEN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015001.2 | NP_055816.2 | 6 - 7 | 1600 | 76 | |
| GenBank mRNA | AF356524.1 | - | 6 - 7 | 1600 | 76 | |
| AK092332.1 | - | 2 - 3 | 173 | 76 | ||
| AK127577.1 | - | 4 - 5 | 923 | 76 | ||
| AL096858.1 | - | 3 - 4 | 518 | 76 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000198.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
mRNA splicing, via spliceosome
transcription, DNA-templated
Notch signaling pathway
viral process
negative regulation of transcription, DNA-templated
positive regulation of neurogenesis
nucleotide binding
RNA polymerase II transcription factor binding
transcriptional repressor activity, RNA polymerase II transcription factor binding
nucleic acid binding
DNA binding
protein binding
poly(A) RNA binding
negative regulation of transcription from RNA polymerase II promoter
mRNA splicing, via spliceosome
transcription, DNA-templated
Notch signaling pathway
viral process
negative regulation of transcription, DNA-templated
positive regulation of neurogenesis
nucleotide binding
RNA polymerase II transcription factor binding
transcriptional repressor activity, RNA polymerase II transcription factor binding
nucleic acid binding
DNA binding
protein binding
poly(A) RNA binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100007006.hg.1 | ENST00000375759.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007006.hg.1 | ENST00000375759.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007006.hg.1 | ENST00000375759.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007006.hg.1 | ENST00000375759.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007006.hg.1 | ENST00000375759.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007006.hg.1 | ENST00000375759.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007006.hg.1 | ENST00000375759.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007006.hg.1 | ENST00000375759.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007006.hg.1 | ENST00000375759.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007006.hg.1 | ENST00000375759.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007006.hg.1 | ENST00000375759.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007006.hg.1 | ENST00000375759.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007006.hg.1 | ENST00000375759.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007006.hg.1 | ENST00000375759.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007006.hg.1 | ENST00000375759.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007006.hg.1 | ENST00000375759.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007006.hg.1 | ENST00000375759.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007006.hg.1 | ENST00000375759.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007006.hg.1 | ENST00000375759.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007006.hg.1 | ENST00000375759.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007006.hg.1 | ENST00000438066.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007006.hg.1 | ENST00000438066.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007006.hg.1 | ENST00000438066.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007006.hg.1 | ENST00000438066.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007006.hg.1 | ENST00000438066.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007006.hg.1 | ENST00000438066.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007006.hg.1 | ENST00000442985.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007006.hg.1 | ENST00000442985.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007006.hg.1 | ENST00000442985.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007006.hg.1 | ENST00000442985.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007006.hg.1 | ENST00000442985.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007006.hg.1 | ENST00000442985.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007006.hg.1 | ENST00000442985.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007006.hg.1 | ENST00000471538.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007006.hg.1 | ENST00000471538.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007006.hg.1 | ENST00000471538.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007006.hg.1 | ENST00000471538.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007006.hg.1 | ENST00000471538.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007006.hg.1 | ENST00000487496.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007006.hg.1 | ENST00000487496.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007006.hg.1 | ENST00000487496.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007006.hg.1 | ENST00000487496.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007006.hg.1 | NM_015001.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007006.hg.1 | NM_015001.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007006.hg.1 | NM_015001.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007006.hg.1 | NM_015001.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007006.hg.1 | NM_015001.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007006.hg.1 | NM_015001.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007006.hg.1 | NM_015001.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007006.hg.1 | NM_015001.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007006.hg.1 | NM_015001.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007006.hg.1 | NM_015001.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007006.hg.1 | NM_015001.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007006.hg.1 | NM_015001.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007006.hg.1 | NM_015001.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007006.hg.1 | NM_015001.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007006.hg.1 | NM_015001.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007006.hg.1 | NM_015001.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007006.hg.1 | NM_015001.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007006.hg.1 | NM_015001.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007006.hg.1 | NM_015001.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007006.hg.1 | NM_015001.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01003307.hg.1 | TC01000198.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01003307.hg.1 | ENST00000375753.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01003307.hg.1 | ENST00000375759.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01003307.hg.1 | NM_015001.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007006.hg.1 | ENST00000375759.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007006.hg.1 | ENST00000375759.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007006.hg.1 | ENST00000375759.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007006.hg.1 | ENST00000375759.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007006.hg.1 | ENST00000375759.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007006.hg.1 | ENST00000375759.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007006.hg.1 | ENST00000375759.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007006.hg.1 | ENST00000375759.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007006.hg.1 | ENST00000375759.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007006.hg.1 | ENST00000375759.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007006.hg.1 | ENST00000375759.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007006.hg.1 | ENST00000375759.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007006.hg.1 | ENST00000375759.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007006.hg.1 | ENST00000375759.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007006.hg.1 | ENST00000375759.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007006.hg.1 | ENST00000375759.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007006.hg.1 | ENST00000375759.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007006.hg.1 | ENST00000375759.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007006.hg.1 | ENST00000375759.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007006.hg.1 | ENST00000375759.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007006.hg.1 | ENST00000438066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007006.hg.1 | ENST00000438066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007006.hg.1 | ENST00000438066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007006.hg.1 | ENST00000438066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007006.hg.1 | ENST00000438066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007006.hg.1 | ENST00000438066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007006.hg.1 | ENST00000442985.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007006.hg.1 | ENST00000442985.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007006.hg.1 | ENST00000442985.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007006.hg.1 | ENST00000442985.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007006.hg.1 | ENST00000442985.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007006.hg.1 | ENST00000442985.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007006.hg.1 | ENST00000442985.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007006.hg.1 | ENST00000471538.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007006.hg.1 | ENST00000471538.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007006.hg.1 | ENST00000471538.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007006.hg.1 | ENST00000471538.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007006.hg.1 | ENST00000471538.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007006.hg.1 | ENST00000487496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007006.hg.1 | ENST00000487496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007006.hg.1 | ENST00000487496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007006.hg.1 | ENST00000487496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007006.hg.1 | NM_015001.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007006.hg.1 | NM_015001.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007006.hg.1 | NM_015001.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007006.hg.1 | NM_015001.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007006.hg.1 | NM_015001.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007006.hg.1 | NM_015001.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007006.hg.1 | NM_015001.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007006.hg.1 | NM_015001.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007006.hg.1 | NM_015001.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007006.hg.1 | NM_015001.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007006.hg.1 | NM_015001.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007006.hg.1 | NM_015001.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007006.hg.1 | NM_015001.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007006.hg.1 | NM_015001.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007006.hg.1 | NM_015001.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007006.hg.1 | NM_015001.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007006.hg.1 | NM_015001.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007006.hg.1 | NM_015001.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007006.hg.1 | NM_015001.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007006.hg.1 | NM_015001.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100148930.hg.1 | ENST00000375759.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100148930.hg.1 | ENST00000442985.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100148930.hg.1 | NM_015001.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100148930.hg.1 | ENST00000375759.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100148930.hg.1 | ENST00000442985.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100148930.hg.1 | NM_015001.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100148930.hg.1 | TC0100007006.hg.1 | Clariom D TC |

SPEN
Genomic Map