Assay Details
| Gene Symbol: RBP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001323518.1 | NP_001310447.1 | 1 - 2 | 75 | 61 | |
More Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
retinoid metabolic process
eye development
gluconeogenesis
heart development
visual perception
maintenance of gastrointestinal epithelium
lung development
positive regulation of insulin secretion
response to retinoic acid
retinol transport
retinol metabolic process
glucose homeostasis
response to ethanol
embryonic organ morphogenesis
embryonic skeletal system development
cardiac muscle tissue development
female genitalia morphogenesis
positive regulation of immunoglobulin secretion
negative regulation of cardiac muscle cell proliferation
embryonic retina morphogenesis in camera-type eye
uterus development
vagina development
urinary bladder development
heart trabecula formation
protein binding
retinal binding
retinol binding
retinol transporter activity
protein heterodimerization activity
retinoid metabolic process
eye development
gluconeogenesis
heart development
visual perception
maintenance of gastrointestinal epithelium
lung development
positive regulation of insulin secretion
response to retinoic acid
retinol transport
retinol metabolic process
glucose homeostasis
response to ethanol
embryonic organ morphogenesis
embryonic skeletal system development
cardiac muscle tissue development
female genitalia morphogenesis
positive regulation of immunoglobulin secretion
negative regulation of cardiac muscle cell proliferation
embryonic retina morphogenesis in camera-type eye
uterus development
vagina development
urinary bladder development
heart trabecula formation
protein binding
retinal binding
retinol binding
retinol transporter activity
protein heterodimerization activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000011450.hg.1 | ENST00000371464.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011450.hg.1 | ENST00000371464.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011450.hg.1 | ENST00000371464.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011450.hg.1 | ENST00000371464.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011450.hg.1 | ENST00000371464.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011450.hg.1 | ENST00000371464.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011450.hg.1 | ENST00000371464.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011450.hg.1 | ENST00000371464.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011450.hg.1 | ENST00000371464.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011450.hg.1 | ENST00000371464.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011450.hg.1 | ENST00000371467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011450.hg.1 | ENST00000371467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011450.hg.1 | ENST00000371467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011450.hg.1 | ENST00000371467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011450.hg.1 | ENST00000371467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011450.hg.1 | ENST00000371467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011450.hg.1 | ENST00000371467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011450.hg.1 | ENST00000371467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011450.hg.1 | ENST00000371467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011450.hg.1 | ENST00000371467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011450.hg.1 | ENST00000371469.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011450.hg.1 | ENST00000371469.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011450.hg.1 | ENST00000371469.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011450.hg.1 | ENST00000371469.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011450.hg.1 | ENST00000371469.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011450.hg.1 | ENST00000371469.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011450.hg.1 | ENST00000371469.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011450.hg.1 | ENST00000371469.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011450.hg.1 | ENST00000615669.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011450.hg.1 | ENST00000615669.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011450.hg.1 | ENST00000615669.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011450.hg.1 | ENST00000615669.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011450.hg.1 | ENST00000615669.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011450.hg.1 | ENST00000615669.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011450.hg.1 | ENST00000615669.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011450.hg.1 | ENST00000615669.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011450.hg.1 | ENST00000615669.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011450.hg.1 | ENST00000629763.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011450.hg.1 | ENST00000629763.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011450.hg.1 | ENST00000629763.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011450.hg.1 | ENST00000629763.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011450.hg.1 | ENST00000629763.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011450.hg.1 | ENST00000629763.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011450.hg.1 | ENST00000629763.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011450.hg.1 | ENST00000629763.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011450.hg.1 | NM_006744.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011450.hg.1 | NM_006744.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011450.hg.1 | NM_006744.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011450.hg.1 | NM_006744.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011450.hg.1 | NM_006744.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011450.hg.1 | NM_006744.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011450.hg.1 | NM_006744.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011450.hg.1 | NM_006744.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011450.hg.1 | NM_006744.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011450.hg.1 | ENST00000371464.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011450.hg.1 | ENST00000371464.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011450.hg.1 | ENST00000371464.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011450.hg.1 | ENST00000371464.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011450.hg.1 | ENST00000371464.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011450.hg.1 | ENST00000371464.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011450.hg.1 | ENST00000371464.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011450.hg.1 | ENST00000371464.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011450.hg.1 | ENST00000371464.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011450.hg.1 | ENST00000371464.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011450.hg.1 | ENST00000371467.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011450.hg.1 | ENST00000371467.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011450.hg.1 | ENST00000371467.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011450.hg.1 | ENST00000371467.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011450.hg.1 | ENST00000371467.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011450.hg.1 | ENST00000371467.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011450.hg.1 | ENST00000371467.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011450.hg.1 | ENST00000371467.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011450.hg.1 | ENST00000371467.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011450.hg.1 | ENST00000371467.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011450.hg.1 | ENST00000371469.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011450.hg.1 | ENST00000371469.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011450.hg.1 | ENST00000371469.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011450.hg.1 | ENST00000371469.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011450.hg.1 | ENST00000371469.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011450.hg.1 | ENST00000371469.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011450.hg.1 | ENST00000371469.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011450.hg.1 | ENST00000371469.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011450.hg.1 | ENST00000615669.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011450.hg.1 | ENST00000615669.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011450.hg.1 | ENST00000615669.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011450.hg.1 | ENST00000615669.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011450.hg.1 | ENST00000615669.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011450.hg.1 | ENST00000615669.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011450.hg.1 | ENST00000615669.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011450.hg.1 | ENST00000615669.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011450.hg.1 | ENST00000615669.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011450.hg.1 | ENST00000629763.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011450.hg.1 | ENST00000629763.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011450.hg.1 | ENST00000629763.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011450.hg.1 | ENST00000629763.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011450.hg.1 | ENST00000629763.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011450.hg.1 | ENST00000629763.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011450.hg.1 | ENST00000629763.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011450.hg.1 | ENST00000629763.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011450.hg.1 | NM_006744.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011450.hg.1 | NM_006744.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011450.hg.1 | NM_006744.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011450.hg.1 | NM_006744.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011450.hg.1 | NM_006744.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011450.hg.1 | NM_006744.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011450.hg.1 | NM_006744.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011450.hg.1 | NM_006744.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011450.hg.1 | NM_006744.3 | Refseq |

RBP4
Genomic Map