Assay Details
| Gene Symbol: PDLIM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020992.3 | NP_066272.1 | 3 - 4 | 568 | 73 | |
| XM_011540330.1 | XP_011538632.1 | 2 - 3 | 274 | 73 | ||
| GenBank mRNA | AJ310549.1 | - | 3 - 4 | 335 | 73 | |
| AK314792.1 | - | 3 - 4 | 425 | 73 | ||
| BC000915.2 | - | 3 - 4 | 444 | 2985229 | 73 | |
| BC018755.1 | - | 3 - 4 | 359 | 3610938 | 73 | |
| FJ224331.1 | - | 3 - 4 | 359 | 73 | ||
| U90878.1 | - | 3 - 4 | 477 | 73 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011540330.1 transcripts.
Set Membership: |
|

PDLIM1
Genomic Map