Assay Details
| Gene Symbol: LRMP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204126.1 | NP_001191055.1 | 10 - 11 | 1452 | 86 | |
| NM_001204127.1 | NP_001191056.1 | 10 - 11 | 1382 | 86 | ||
| NM_006152.3 | NP_006143.2 | 11 - 12 | 1533 | 86 | ||
| XM_005253370.3 | XP_005253427.1 | 12 - 13 | 1592 | 86 | ||
| XM_005253372.2 | XP_005253429.1 | 11 - 12 | 1593 | 86 | ||
| XM_005253374.3 | XP_005253431.1 | 9 - 10 | 1450 | 86 | ||
| XM_006719076.2 | XP_006719139.1 | 8 - 9 | 516 | 86 | ||
| XM_011520667.1 | XP_011518969.1 | 12 - 13 | 1741 | 86 | ||
| XM_011520668.2 | XP_011518970.1 | 8 - 9 | 779 | 86 | ||
| XM_011520669.2 | XP_011518971.1 | 7 - 8 | 661 | 86 | ||
| XM_011520670.1 | XP_011518972.1 | 7 - 8 | 432 | 86 | ||
| XM_017019300.1 | XP_016874789.1 | 11 - 12 | 1676 | 86 | ||
| XM_017019301.1 | XP_016874790.1 | 10 - 11 | 1385 | 86 | ||
| XM_017019302.1 | XP_016874791.1 | 11 - 12 | 1590 | 86 | ||
| GenBank mRNA | AK303103.1 | - | 7 - 8 | 785 | 86 | |
| AK303256.1 | - | 10 - 11 | 991 | 86 | ||
| BC126417.1 | - | 11 - 12 | 1040 | 8992133 | 86 | |
| BC144164.1 | - | 10 - 11 | 959 | 86 | ||
| BC144165.1 | - | 11 - 12 | 1040 | 86 | ||
| U10485.1 | - | 11 - 12 | 1181 | 86 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the XM_011520670.1 transcripts.
Set Membership: |
|

LRMP
Genomic Map