Assay Details
| Gene Symbol: LRMP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005253370.3 | XP_005253427.1 | 2 - 3 | 767 | 128 | |
| XM_005253372.2 | XP_005253429.1 | 2 - 3 | 913 | 128 | ||
| XM_011520667.1 | XP_011518969.1 | 2 - 3 | 913 | 128 | ||
| XM_017019302.1 | XP_016874791.1 | 2 - 3 | 913 | 128 | ||
| GenBank mRNA | BC029391.1 | - | 2 - 3 | 450 | 128 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005253370.3, XM_005253372.2, XM_011520667.1, XM_017019302.1 transcripts.
Set Membership: |
|

LRMP
Genomic Map