Assay Details
| Gene Symbol: FANCG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK293427.1 | - | 1 - 2 | 169 | 67 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK293427.1 transcripts.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | ENST00000378643.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | ENST00000378643.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | ENST00000378643.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | ENST00000378643.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | ENST00000378643.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | ENST00000378643.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | ENST00000378643.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | ENST00000378643.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | ENST00000378643.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | ENST00000378643.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | ENST00000378643.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | ENST00000378643.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | ENST00000378643.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | ENST00000378643.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | ENST00000378643.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | ENST00000378643.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | ENST00000378643.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | ENST00000378643.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | ENST00000378643.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | ENST00000378643.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | ENST00000378643.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | ENST00000378643.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | ENST00000378643.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | ENST00000378643.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | ENST00000425676.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | ENST00000425676.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | ENST00000425676.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | ENST00000425676.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | ENST00000425676.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | ENST00000425676.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | ENST00000425676.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | ENST00000425676.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | ENST00000425676.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | ENST00000425676.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | ENST00000425676.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | ENST00000425676.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | ENST00000425676.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | ENST00000425676.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | ENST00000425676.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | ENST00000425676.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | ENST00000425676.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | ENST00000425676.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | ENST00000425676.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | ENST00000448890.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | ENST00000448890.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | ENST00000448890.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | ENST00000448890.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | ENST00000448890.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | ENST00000448890.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | ENST00000448890.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | ENST00000448890.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | ENST00000461149.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | ENST00000461149.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | ENST00000461149.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | ENST00000462124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | ENST00000462124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | ENST00000462124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | ENST00000462124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | ENST00000462124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | ENST00000462124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | ENST00000474894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | ENST00000474894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | ENST00000474894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | ENST00000474894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | ENST00000476212.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | ENST00000476212.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | ENST00000476212.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | ENST00000476212.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | ENST00000481254.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | ENST00000481254.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | ENST00000481254.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | ENST00000481254.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | ENST00000481254.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | NM_004629.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | NM_004629.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | NM_004629.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | NM_004629.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | NM_004629.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | NM_004629.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | NM_004629.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | NM_004629.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | NM_004629.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | NM_004629.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | NM_004629.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | NM_004629.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | NM_004629.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | NM_004629.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | NM_004629.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | NM_004629.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | NM_004629.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | NM_004629.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | NM_004629.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | NM_004629.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | NM_004629.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | NM_004629.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | NM_004629.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009930.hg.1 | NM_004629.1 | Refseq |

FANCG
Genomic Map