Assay Details
| Gene Symbol: GABARAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007278.1 | NP_009209.1 | 4 | 460 | 111 | |
| GenBank mRNA | AB030711.1 | - | 4 | 459 | 111 | |
| AF044671.1 | - | 4 | 434 | 111 | ||
| AF067171.1 | - | 4 | 460 | 111 | ||
| AF161586.1 | - | 4 | 424 | 111 | ||
| AF183425.1 | - | 4 | 459 | 111 | ||
| AK098634.1 | - | 4 | 464 | 111 | ||
| AL050182.1 | - | 366 | 111 | |||
| BC106748.1 | - | 4 | 402 | 40022383 | 111 | |
| BC106749.1 | - | 4 | 402 | 40022384 | 111 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

GABARAP
Genomic Map